Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 56963694-56963967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPAD1
Entrez Gene ID
54928
Gene Name
inositol monophosphatase domain containing 1
Gene Aliases
GPAPP, IMP 3, IMP-3, IMPA3
UniGene
Hs.438689
Gene Chromosome Location
Chr.8: 56957929-56993874 on Build GRCh38
Transcripts
NM_017813.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100687 c.1064A>G p.K355R Chr.8: 56963809-56963809 on Build GRCh38
6685617 c.1023G>T p.M341I Chr.8: 56963850-56963850 on Build GRCh38
6039539 c.939delG p.H314fs*2 Chr.8: 56963934-56963934 on Build GRCh38
403004 c.937G>T p.G313W Chr.8: 56963936-56963936 on Build GRCh38
1194272 c.910G>T p.G304C Chr.8: 56963963-56963963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756069211 Chr.8: 56963790-56963790 on Build GRCh38
rs772424020 Chr.8: 56963934-56963934 on Build GRCh38
rs748533711 Chr.8: 56963837-56963837 on Build GRCh38
rs552550687 Chr.8: 56963919-56963919 on Build GRCh38
rs763327486 Chr.8: 56963875-56963875 on Build GRCh38
rs564170685 Chr.8: 56963722-56963722 on Build GRCh38
rs201175538 Chr.8: 56963807-56963807 on Build GRCh38
rs977456990 Chr.8: 56963729-56963729 on Build GRCh38
rs767359786 Chr.8: 56963786-56963786 on Build GRCh38
rs779233560 Chr.8: 56963863-56963864 on Build GRCh38
rs371111058 Chr.8: 56963723-56963723 on Build GRCh38
rs770253673 Chr.8: 56963839-56963839 on Build GRCh38
rs376844118 Chr.8: 56963787-56963787 on Build GRCh38
rs112201304 Chr.8: 56963934-56963934 on Build GRCh38
rs745979731 Chr.8: 56963802-56963802 on Build GRCh38
rs925947188 Chr.8: 56963736-56963736 on Build GRCh38
rs76235334 Chr.8: 56963909-56963909 on Build GRCh38
rs762476811 Chr.8: 56963771-56963771 on Build GRCh38
rs560353101 Chr.8: 56963819-56963819 on Build GRCh38
rs752543382 Chr.8: 56963898-56963898 on Build GRCh38
rs765934902 Chr.8: 56963882-56963882 on Build GRCh38
rs768948674 Chr.8: 56963773-56963774 on Build GRCh38
rs992299291 Chr.8: 56963725-56963725 on Build GRCh38
rs772611352 Chr.8: 56963758-56963758 on Build GRCh38
rs372925433 Chr.8: 56963832-56963832 on Build GRCh38
rs546293123 Chr.8: 56963800-56963800 on Build GRCh38
rs906499247 Chr.8: 56963838-56963838 on Build GRCh38
rs1005277199 Chr.8: 56963905-56963905 on Build GRCh38
rs780287418 Chr.8: 56963937-56963937 on Build GRCh38
rs751254735 Chr.8: 56963781-56963781 on Build GRCh38
rs748575076 Chr.8: 56963871-56963871 on Build GRCh38
rs34467829 Chr.8: 56963886-56963886 on Build GRCh38
rs922875995 Chr.8: 56963721-56963721 on Build GRCh38
rs768934175 Chr.8: 56963829-56963829 on Build GRCh38
rs375887598 Chr.8: 56963874-56963874 on Build GRCh38
rs763761420 Chr.8: 56963917-56963917 on Build GRCh38
rs370198289 Chr.8: 56963938-56963938 on Build GRCh38
rs765979313 Chr.8: 56963778-56963778 on Build GRCh38
rs918107288 Chr.8: 56963777-56963777 on Build GRCh38
rs752559453 Chr.8: 56963738-56963738 on Build GRCh38
rs755310573 Chr.8: 56963939-56963939 on Build GRCh38
rs1038937444 Chr.8: 56963821-56963821 on Build GRCh38
rs765707549 Chr.8: 56963745-56963745 on Build GRCh38
rs747249859 Chr.8: 56963820-56963820 on Build GRCh38
rs773790200 Chr.8: 56963884-56963884 on Build GRCh38
rs886063018 Chr.8: 56963779-56963779 on Build GRCh38
rs1030083794 Chr.8: 56963841-56963841 on Build GRCh38
rs781542516 Chr.8: 56963942-56963942 on Build GRCh38
rs138502852 Chr.8: 56963885-56963885 on Build GRCh38

View Full Product Details