Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30463176-30463449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGB1
Entrez Gene ID
3146
Gene Name
high mobility group box 1
Gene Aliases
HMG-1, HMG1, HMG3, SBP-1
UniGene
Hs.434102
Gene Chromosome Location
Chr.13: 30457916-30617597 on Build GRCh38
Transcripts
NM_001313892.1 NM_001313893.1 NM_002128.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5655361 c.151-2A>T p.? Chr.13: 30463354-30463354 on Build GRCh38
6336690 c.195A>G p.K65K Chr.13: 30463308-30463308 on Build GRCh38
3793236 c.166G>C p.E56Q Chr.13: 30463337-30463337 on Build GRCh38
6388023 c.289A>G p.R97G Chr.13: 30463214-30463214 on Build GRCh38
946621 c.253A>G p.T85A Chr.13: 30463250-30463250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777739337 Chr.13: 30463368-30463368 on Build GRCh38
rs371450041 Chr.13: 30463387-30463387 on Build GRCh38
rs143387796 Chr.13: 30463233-30463233 on Build GRCh38
rs2388544 Chr.13: 30463210-30463210 on Build GRCh38
rs760440175 Chr.13: 30463212-30463212 on Build GRCh38
rs1048590358 Chr.13: 30463388-30463388 on Build GRCh38
rs780768302 Chr.13: 30463382-30463382 on Build GRCh38
rs983723 Chr.13: 30463308-30463308 on Build GRCh38
rs746817748 Chr.13: 30463380-30463380 on Build GRCh38
rs372814279 Chr.13: 30463423-30463423 on Build GRCh38
rs756214554 Chr.13: 30463262-30463262 on Build GRCh38
rs758359066 Chr.13: 30463326-30463326 on Build GRCh38
rs780215433 Chr.13: 30463269-30463269 on Build GRCh38
rs367581777 Chr.13: 30463381-30463381 on Build GRCh38
rs912774039 Chr.13: 30463305-30463305 on Build GRCh38
rs750030870 Chr.13: 30463388-30463391 on Build GRCh38
rs767679881 Chr.13: 30463227-30463227 on Build GRCh38
rs1025665018 Chr.13: 30463412-30463412 on Build GRCh38
rs866251338 Chr.13: 30463421-30463421 on Build GRCh38
rs375874287 Chr.13: 30463370-30463370 on Build GRCh38
rs956895154 Chr.13: 30463248-30463248 on Build GRCh38
rs988415728 Chr.13: 30463290-30463290 on Build GRCh38
rs762127662 Chr.13: 30463224-30463224 on Build GRCh38
rs908220823 Chr.13: 30463399-30463399 on Build GRCh38
rs11546137 Chr.13: 30463283-30463283 on Build GRCh38
rs193056442 Chr.13: 30463383-30463383 on Build GRCh38

View Full Product Details