Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38414528-38414781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1
Entrez Gene ID
2260
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
UniGene
Hs.264887
Gene Chromosome Location
Chr.8: 38411138-38468834 on Build GRCh38
Transcripts
NM_001174063.1 NM_001174064.1 NM_001174065.1 NM_001174066.1 NM_001174067.1 NM_015850.3 NM_023105.2 NM_023106.2 NM_023110.2 XM_006716303.2 XM_006716304.1 XM_006716306.2 XM_006716307.1 XM_006716309.3 XM_006716310.2 XM_006716311.1 XM_006716312.1 XM_006716313.2 XM_006716314.1 XM_011544443.2 XM_011544444.1 XM_011544445.2 XM_011544446.2 XM_011544447.2 XM_011544448.1 XM_011544449.1 XM_011544450.2 XM_011544451.1 XM_017013219.1 XM_017013220.1 XM_017013221.1 XM_017013222.1 XM_017013223.1 XM_017013224.1 XM_017013225.1 XM_017013226.1 XM_017013227.1 XM_017013228.1 XM_017013229.1 XM_017013230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4767725 c.1947C>T p.N649N Chr.8: 38414779-38414779 on Build GRCh38
6964736 c.1982G>A p.R661Q Chr.8: 38414625-38414625 on Build GRCh38
5874653 c.1948G>A p.G650S Chr.8: 38414629-38414629 on Build GRCh38
6964735 c.1982G>A p.R661Q Chr.8: 38414625-38414625 on Build GRCh38
1207083 c.2024G>A p.R675Q Chr.8: 38414583-38414583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764971696 Chr.8: 38414645-38414645 on Build GRCh38
rs1057518620 Chr.8: 38414617-38414617 on Build GRCh38
rs779120005 Chr.8: 38414749-38414749 on Build GRCh38
rs1029181940 Chr.8: 38414728-38414728 on Build GRCh38
rs145273148 Chr.8: 38414739-38414739 on Build GRCh38
rs746473852 Chr.8: 38414723-38414723 on Build GRCh38
rs397515446 Chr.8: 38414599-38414599 on Build GRCh38
rs983971417 Chr.8: 38414699-38414699 on Build GRCh38
rs758306938 Chr.8: 38414628-38414629 on Build GRCh38
rs927214668 Chr.8: 38414736-38414736 on Build GRCh38
rs751129940 Chr.8: 38414665-38414665 on Build GRCh38
rs2978072 Chr.8: 38414685-38414685 on Build GRCh38
rs778015347 Chr.8: 38414732-38414732 on Build GRCh38
rs755456709 Chr.8: 38414670-38414670 on Build GRCh38
rs950907705 Chr.8: 38414663-38414663 on Build GRCh38
rs776590352 Chr.8: 38414760-38414760 on Build GRCh38
rs761708658 Chr.8: 38414642-38414642 on Build GRCh38
rs148750337 Chr.8: 38414763-38414763 on Build GRCh38
rs749546240 Chr.8: 38414561-38414561 on Build GRCh38
rs756455400 Chr.8: 38414681-38414681 on Build GRCh38
rs917323541 Chr.8: 38414701-38414701 on Build GRCh38
rs750042204 Chr.8: 38414646-38414646 on Build GRCh38
rs201846022 Chr.8: 38414651-38414651 on Build GRCh38
rs771078736 Chr.8: 38414583-38414583 on Build GRCh38
rs869217053 Chr.8: 38414724-38414725 on Build GRCh38
rs781745049 Chr.8: 38414672-38414672 on Build GRCh38
rs112311314 Chr.8: 38414637-38414637 on Build GRCh38
rs201805975 Chr.8: 38414761-38414761 on Build GRCh38
rs869281958 Chr.8: 38414635-38414635 on Build GRCh38
rs749452174 Chr.8: 38414741-38414741 on Build GRCh38
rs745989679 Chr.8: 38414752-38414752 on Build GRCh38
rs765870661 Chr.8: 38414654-38414654 on Build GRCh38
rs375611478 Chr.8: 38414584-38414584 on Build GRCh38
rs199601282 Chr.8: 38414679-38414679 on Build GRCh38
rs121909636 Chr.8: 38414569-38414569 on Build GRCh38
rs768446923 Chr.8: 38414757-38414757 on Build GRCh38
rs760681522 Chr.8: 38414600-38414600 on Build GRCh38
rs771342533 Chr.8: 38414746-38414746 on Build GRCh38
rs776264072 Chr.8: 38414626-38414626 on Build GRCh38

View Full Product Details