Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216694626-216694851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFBP5
Entrez Gene ID
3488
Gene Name
insulin like growth factor binding protein 5
Gene Aliases
IBP5
UniGene
Hs.607212
Gene Chromosome Location
Chr.2: 216672105-216695549 on Build GRCh38
Transcripts
NM_000599.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7196090 c.41A>G p.Y14C Chr.2: 216694735-216694735 on Build GRCh38
6684335 c.29T>C p.L10P Chr.2: 216694747-216694747 on Build GRCh38
5956952 c.63G>C p.L21L Chr.2: 216694713-216694713 on Build GRCh38
4091345 c.37G>A p.A13T Chr.2: 216694739-216694739 on Build GRCh38
4613560 c.120delC p.S41fs*17 Chr.2: 216694656-216694656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763488205 Chr.2: 216694730-216694730 on Build GRCh38
rs1031161465 Chr.2: 216694828-216694828 on Build GRCh38
rs929724774 Chr.2: 216694682-216694682 on Build GRCh38
rs202117764 Chr.2: 216694713-216694713 on Build GRCh38
rs771103319 Chr.2: 216694656-216694656 on Build GRCh38
rs759992658 Chr.2: 216694724-216694724 on Build GRCh38
rs764132504 Chr.2: 216694731-216694731 on Build GRCh38
rs748069601 Chr.2: 216694697-216694697 on Build GRCh38
rs930571753 Chr.2: 216694656-216694656 on Build GRCh38
rs772834319 Chr.2: 216694685-216694685 on Build GRCh38
rs867462332 Chr.2: 216694739-216694739 on Build GRCh38
rs775897614 Chr.2: 216694726-216694726 on Build GRCh38
rs764197301 Chr.2: 216694653-216694653 on Build GRCh38
rs754697428 Chr.2: 216694681-216694681 on Build GRCh38
rs1000194504 Chr.2: 216694821-216694821 on Build GRCh38
rs758433852 Chr.2: 216694652-216694652 on Build GRCh38
rs753202179 Chr.2: 216694650-216694650 on Build GRCh38
rs191919473 Chr.2: 216694758-216694758 on Build GRCh38
rs1047516241 Chr.2: 216694658-216694658 on Build GRCh38
rs865878064 Chr.2: 216694715-216694715 on Build GRCh38
rs771512778 Chr.2: 216694699-216694699 on Build GRCh38
rs11575124 Chr.2: 216694779-216694779 on Build GRCh38
rs765651842 Chr.2: 216694725-216694725 on Build GRCh38
rs1012231136 Chr.2: 216694805-216694805 on Build GRCh38
rs772745562 Chr.2: 216694700-216694700 on Build GRCh38
rs968290360 Chr.2: 216694820-216694820 on Build GRCh38
rs765843936 Chr.2: 216694646-216694646 on Build GRCh38
rs750582410 Chr.2: 216694789-216694789 on Build GRCh38
rs748087175 Chr.2: 216694655-216694656 on Build GRCh38
rs749074136 Chr.2: 216694662-216694662 on Build GRCh38
rs867069708 Chr.2: 216694727-216694727 on Build GRCh38
rs776355825 Chr.2: 216694717-216694717 on Build GRCh38
rs778834706 Chr.2: 216694684-216694684 on Build GRCh38
rs564421832 Chr.2: 216694711-216694711 on Build GRCh38
rs1022271956 Chr.2: 216694807-216694807 on Build GRCh38
rs757476772 Chr.2: 216694659-216694659 on Build GRCh38
rs751642194 Chr.2: 216694740-216694740 on Build GRCh38
rs767727294 Chr.2: 216694787-216694787 on Build GRCh38
rs779879411 Chr.2: 216694661-216694661 on Build GRCh38
rs1046660209 Chr.2: 216694647-216694647 on Build GRCh38
rs763336377 Chr.2: 216694791-216694791 on Build GRCh38

View Full Product Details