Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139398516-139398755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf55-LUC7L2
Entrez Gene ID
100996928
Gene Name
C7orf55-LUC7L2 readthrough
Gene Aliases
-
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139341132-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_001244585.1 NM_001270643.1 NM_016019.4

Gene Symbol
LUC7L2
Entrez Gene ID
51631
Gene Name
LUC7 like 2, pre-mRNA splicing factor
Gene Aliases
CGI-59, CGI-74, LUC7B2
UniGene
Hs.731488
Gene Chromosome Location
Chr.7: 139340359-139423457 on Build GRCh38
Transcripts
NM_001244584.2 NM_001244585.1 NM_001270643.1 NM_016019.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5195985 c.251_252+2delATGT p.? Chr.7: 139398696-139398699 on Build GRCh38
4994073 c.214G>T p.E72* Chr.7: 139398656-139398656 on Build GRCh38
7088499 c.177G>A p.L59L Chr.7: 139398622-139398622 on Build GRCh38
4994074 c.211G>T p.E71* Chr.7: 139398656-139398656 on Build GRCh38
5830588 c.213A>C p.E71D Chr.7: 139398658-139398658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000583962 Chr.7: 139398622-139398622 on Build GRCh38
rs768500321 Chr.7: 139398553-139398553 on Build GRCh38
rs200748270 Chr.7: 139398703-139398703 on Build GRCh38
rs777999061 Chr.7: 139398649-139398649 on Build GRCh38
rs1019128511 Chr.7: 139398714-139398714 on Build GRCh38
rs751873571 Chr.7: 139398655-139398655 on Build GRCh38
rs746086729 Chr.7: 139398552-139398553 on Build GRCh38
rs199947447 Chr.7: 139398692-139398692 on Build GRCh38
rs754174949 Chr.7: 139398579-139398579 on Build GRCh38
rs761235355 Chr.7: 139398560-139398560 on Build GRCh38
rs776925357 Chr.7: 139398553-139398553 on Build GRCh38
rs202002472 Chr.7: 139398574-139398574 on Build GRCh38
rs754735833 Chr.7: 139398701-139398701 on Build GRCh38
rs771514427 Chr.7: 139398568-139398568 on Build GRCh38
rs759498547 Chr.7: 139398571-139398576 on Build GRCh38
rs528492867 Chr.7: 139398634-139398634 on Build GRCh38
rs773261565 Chr.7: 139398584-139398584 on Build GRCh38
rs749300728 Chr.7: 139398552-139398552 on Build GRCh38
rs1033293553 Chr.7: 139398645-139398645 on Build GRCh38
rs766778920 Chr.7: 139398575-139398575 on Build GRCh38
rs368837024 Chr.7: 139398582-139398582 on Build GRCh38
rs373024282 Chr.7: 139398610-139398610 on Build GRCh38
rs757535398 Chr.7: 139398659-139398659 on Build GRCh38
rs769848355 Chr.7: 139398559-139398560 on Build GRCh38
rs966168744 Chr.7: 139398723-139398723 on Build GRCh38
rs781430424 Chr.7: 139398661-139398661 on Build GRCh38
rs765662894 Chr.7: 139398601-139398601 on Build GRCh38
rs772914516 Chr.7: 139398573-139398573 on Build GRCh38
rs775783093 Chr.7: 139398560-139398560 on Build GRCh38
rs762901221 Chr.7: 139398560-139398563 on Build GRCh38
rs748110774 Chr.7: 139398718-139398718 on Build GRCh38
rs907675494 Chr.7: 139398662-139398662 on Build GRCh38
rs760734529 Chr.7: 139398721-139398723 on Build GRCh38
rs797010906 Chr.7: 139398583-139398583 on Build GRCh38

View Full Product Details