Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11649100-11649325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO2
Entrez Gene ID
26232
Gene Name
F-box protein 2
Gene Aliases
FBG1, FBX2, Fbs1, NFB42, OCP1
UniGene
Hs.132753
Gene Chromosome Location
Chr.1: 11648361-11654831 on Build GRCh38
Transcripts
NM_012168.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747659 c.682G>A p.E228K Chr.1: 11649161-11649161 on Build GRCh38
320379 c.700T>A p.F234I Chr.1: 11649143-11649143 on Build GRCh38
894404 c.686A>C p.N229T Chr.1: 11649157-11649157 on Build GRCh38
5016808 c.651C>G p.Y217* Chr.1: 11649192-11649192 on Build GRCh38
6372304 c.649T>A p.Y217N Chr.1: 11649194-11649194 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756719894 Chr.1: 11649260-11649260 on Build GRCh38
rs746580239 Chr.1: 11649162-11649162 on Build GRCh38
rs575386507 Chr.1: 11649147-11649147 on Build GRCh38
rs780609648 Chr.1: 11649244-11649245 on Build GRCh38
rs540294665 Chr.1: 11649245-11649245 on Build GRCh38
rs371864661 Chr.1: 11649184-11649184 on Build GRCh38
rs374850495 Chr.1: 11649206-11649206 on Build GRCh38
rs754041183 Chr.1: 11649212-11649212 on Build GRCh38
rs12725584 Chr.1: 11649278-11649278 on Build GRCh38
rs748063439 Chr.1: 11649152-11649152 on Build GRCh38
rs754760505 Chr.1: 11649140-11649140 on Build GRCh38
rs1013376316 Chr.1: 11649141-11649141 on Build GRCh38
rs200671071 Chr.1: 11649165-11649165 on Build GRCh38
rs907948804 Chr.1: 11649220-11649220 on Build GRCh38
rs745455237 Chr.1: 11649263-11649263 on Build GRCh38
rs762374502 Chr.1: 11649191-11649191 on Build GRCh38
rs573282990 Chr.1: 11649215-11649215 on Build GRCh38
rs758318121 Chr.1: 11649219-11649219 on Build GRCh38
rs1003676942 Chr.1: 11649175-11649175 on Build GRCh38
rs12725708 Chr.1: 11649304-11649304 on Build GRCh38
rs1037850751 Chr.1: 11649250-11649250 on Build GRCh38
rs940863414 Chr.1: 11649232-11649233 on Build GRCh38
rs185490091 Chr.1: 11649153-11649153 on Build GRCh38
rs2272984 Chr.1: 11649282-11649282 on Build GRCh38
rs562080767 Chr.1: 11649261-11649261 on Build GRCh38
rs770552000 Chr.1: 11649167-11649167 on Build GRCh38
rs1032237428 Chr.1: 11649195-11649195 on Build GRCh38
rs147887052 Chr.1: 11649168-11649168 on Build GRCh38
rs746494486 Chr.1: 11649256-11649256 on Build GRCh38
rs879582563 Chr.1: 11649230-11649230 on Build GRCh38
rs774580914 Chr.1: 11649183-11649183 on Build GRCh38
rs753544299 Chr.1: 11649136-11649136 on Build GRCh38
rs965871110 Chr.1: 11649292-11649292 on Build GRCh38
rs868202503 Chr.1: 11649125-11649125 on Build GRCh38
rs768192437 Chr.1: 11649192-11649192 on Build GRCh38
rs12725706 Chr.1: 11649298-11649298 on Build GRCh38
rs774725222 Chr.1: 11649268-11649269 on Build GRCh38
rs749767559 Chr.1: 11649256-11649257 on Build GRCh38
rs769004518 Chr.1: 11649178-11649178 on Build GRCh38
rs766596532 Chr.1: 11649207-11649207 on Build GRCh38
rs367827521 Chr.1: 11649214-11649214 on Build GRCh38
rs777262074 Chr.1: 11649159-11649159 on Build GRCh38

View Full Product Details