Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131274502-131274761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF1
Entrez Gene ID
3547
Gene Name
immunoglobulin superfamily member 1
Gene Aliases
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120
UniGene
Hs.22111
Gene Chromosome Location
Chr.X: 131273506-131289457 on Build GRCh38
Transcripts
NM_001170961.1 NM_001170962.1 NM_001555.4 XM_011531330.1 XM_011531333.1 XM_011531334.1

SNP Detail

SNP ID Chromosome Location
rs772852471 Chr.X: 131274664-131274664 on Build GRCh38
rs186207785 Chr.X: 131274551-131274551 on Build GRCh38
rs778161522 Chr.X: 131274552-131274552 on Build GRCh38
rs745938115 Chr.X: 131274601-131274601 on Build GRCh38
rs148685541 Chr.X: 131274604-131274604 on Build GRCh38
rs771794539 Chr.X: 131274623-131274623 on Build GRCh38
rs760383828 Chr.X: 131274665-131274665 on Build GRCh38
rs1049168036 Chr.X: 131274558-131274558 on Build GRCh38
rs766037546 Chr.X: 131274692-131274692 on Build GRCh38
rs774303182 Chr.X: 131274612-131274612 on Build GRCh38
rs989494756 Chr.X: 131274708-131274708 on Build GRCh38
rs759059710 Chr.X: 131274704-131274704 on Build GRCh38
rs765697743 Chr.X: 131274734-131274734 on Build GRCh38
rs747172056 Chr.X: 131274579-131274579 on Build GRCh38
rs1040781608 Chr.X: 131274524-131274524 on Build GRCh38
rs781196045 Chr.X: 131274582-131274582 on Build GRCh38
rs1023012731 Chr.X: 131274525-131274525 on Build GRCh38
rs868050867 Chr.X: 131274686-131274686 on Build GRCh38
rs757291025 Chr.X: 131274541-131274541 on Build GRCh38
rs761616570 Chr.X: 131274615-131274615 on Build GRCh38
rs138128951 Chr.X: 131274581-131274581 on Build GRCh38
rs377681881 Chr.X: 131274573-131274573 on Build GRCh38
rs753341895 Chr.X: 131274703-131274703 on Build GRCh38

View Full Product Details