Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49847221-49847448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ45513
Entrez Gene ID
729220
Gene Name
uncharacterized LOC729220
Gene Aliases
-
UniGene
Hs.434138
Gene Chromosome Location
Chr.17: 49845910-49848837 on Build GRCh38
Transcripts
NM_001242791.1 NM_001077503.1 NM_001077504.1 NM_001077505.1 NM_001077506.1 NM_170685.2

Gene Symbol
TAC4
Entrez Gene ID
255061
Gene Name
tachykinin 4 (hemokinin)
Gene Aliases
EK, HK-1, HK1, PPT-C
UniGene
Hs.652130
Gene Chromosome Location
Chr.17: 49838309-49848017 on Build GRCh38
Transcripts
NM_001242791.1 NM_001077503.1 NM_001077504.1 NM_001077505.1 NM_001077506.1 NM_170685.2

SNP Detail

SNP ID Chromosome Location
rs780364865 Chr.17: 49847293-49847293 on Build GRCh38
rs918758053 Chr.17: 49847335-49847335 on Build GRCh38
rs962835597 Chr.17: 49847321-49847321 on Build GRCh38
rs371403490 Chr.17: 49847243-49847243 on Build GRCh38
rs188977489 Chr.17: 49847292-49847292 on Build GRCh38
rs756407796 Chr.17: 49847279-49847279 on Build GRCh38
rs779117068 Chr.17: 49847400-49847400 on Build GRCh38
rs749694303 Chr.17: 49847342-49847342 on Build GRCh38
rs983186800 Chr.17: 49847391-49847391 on Build GRCh38
rs748742149 Chr.17: 49847406-49847406 on Build GRCh38
rs779216577 Chr.17: 49847315-49847315 on Build GRCh38
rs977005481 Chr.17: 49847302-49847302 on Build GRCh38
rs536638786 Chr.17: 49847267-49847267 on Build GRCh38
rs765131915 Chr.17: 49847281-49847281 on Build GRCh38
rs770531887 Chr.17: 49847281-49847281 on Build GRCh38
rs755353808 Chr.17: 49847393-49847393 on Build GRCh38
rs1044068903 Chr.17: 49847410-49847410 on Build GRCh38

View Full Product Details