Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47403099-47403358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB1
Entrez Gene ID
2560
Gene Name
gamma-aminobutyric acid type A receptor beta1 subunit
Gene Aliases
-
UniGene
Hs.254117
Gene Chromosome Location
Chr.4: 47031278-47438409 on Build GRCh38
Transcripts
NM_000812.3 XM_017007985.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6290843 c.558T>G p.T186T Chr.4: 47403331-47403331 on Build GRCh38
5866455 c.568G>A p.E190K Chr.4: 47403341-47403341 on Build GRCh38
394799 c.561T>C p.D187D Chr.4: 47403334-47403334 on Build GRCh38
6238590 c.559G>T p.D187Y Chr.4: 47403332-47403332 on Build GRCh38
6668571 c.563A>G p.D188G Chr.4: 47403336-47403336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199730245 Chr.4: 47403305-47403305 on Build GRCh38
rs771361463 Chr.4: 47403176-47403176 on Build GRCh38
rs190472480 Chr.4: 47403172-47403172 on Build GRCh38
rs533587392 Chr.4: 47403247-47403247 on Build GRCh38
rs566541642 Chr.4: 47403216-47403216 on Build GRCh38
rs917611390 Chr.4: 47403198-47403198 on Build GRCh38
rs776407721 Chr.4: 47403277-47403277 on Build GRCh38
rs772264977 Chr.4: 47403275-47403275 on Build GRCh38
rs932147884 Chr.4: 47403149-47403149 on Build GRCh38
rs372855800 Chr.4: 47403296-47403296 on Build GRCh38
rs369649820 Chr.4: 47403302-47403302 on Build GRCh38
rs375958169 Chr.4: 47403299-47403299 on Build GRCh38
rs548028914 Chr.4: 47403191-47403191 on Build GRCh38
rs551195855 Chr.4: 47403156-47403156 on Build GRCh38
rs750749012 Chr.4: 47403307-47403307 on Build GRCh38
rs375002903 Chr.4: 47403287-47403287 on Build GRCh38
rs574247655 Chr.4: 47403239-47403240 on Build GRCh38
rs912187276 Chr.4: 47403196-47403196 on Build GRCh38
rs897642651 Chr.4: 47403306-47403306 on Build GRCh38
rs947714442 Chr.4: 47403219-47403219 on Build GRCh38
rs1055480974 Chr.4: 47403297-47403297 on Build GRCh38
rs762333066 Chr.4: 47403295-47403295 on Build GRCh38
rs924094454 Chr.4: 47403160-47403160 on Build GRCh38
rs756559058 Chr.4: 47403311-47403311 on Build GRCh38
rs200451571 Chr.4: 47403294-47403294 on Build GRCh38
rs773778765 Chr.4: 47403300-47403300 on Build GRCh38
rs774975089 Chr.4: 47403188-47403188 on Build GRCh38
rs145050543 Chr.4: 47403167-47403167 on Build GRCh38

View Full Product Details