Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123059200-123059459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA8
Entrez Gene ID
3312
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
UniGene
Hs.180414
Gene Chromosome Location
Chr.11: 123057492-123062366 on Build GRCh38
Transcripts
NM_006597.5 NM_153201.3 XM_011542798.1 NR_001453.2

Gene Symbol
SNORD14C
Entrez Gene ID
85389
Gene Name
small nucleolar RNA, C/D box 14C
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 123059335-123059422 on Build GRCh38
Transcripts
NM_006597.5 NM_153201.3 XM_011542798.1 NR_001453.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6838140 c.1121C>G p.A374G Chr.11: 123059261-123059261 on Build GRCh38
1210075 c.1171T>G p.L391V Chr.11: 123059211-123059211 on Build GRCh38
1719093 c.1140G>C p.L380F Chr.11: 123059242-123059242 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201521469 Chr.11: 123059400-123059400 on Build GRCh38
rs768379355 Chr.11: 123059304-123059304 on Build GRCh38
rs772121271 Chr.11: 123059242-123059242 on Build GRCh38
rs201838176 Chr.11: 123059420-123059421 on Build GRCh38
rs981966846 Chr.11: 123059300-123059300 on Build GRCh38
rs557485555 Chr.11: 123059342-123059342 on Build GRCh38
rs764411708 Chr.11: 123059392-123059392 on Build GRCh38
rs367639438 Chr.11: 123059264-123059265 on Build GRCh38
rs746781077 Chr.11: 123059261-123059261 on Build GRCh38
rs754070761 Chr.11: 123059393-123059393 on Build GRCh38
rs761439627 Chr.11: 123059352-123059353 on Build GRCh38
rs368360341 Chr.11: 123059278-123059278 on Build GRCh38
rs775435579 Chr.11: 123059257-123059257 on Build GRCh38
rs769407459 Chr.11: 123059285-123059286 on Build GRCh38
rs770428004 Chr.11: 123059366-123059366 on Build GRCh38
rs756399523 Chr.11: 123059288-123059288 on Build GRCh38
rs138440661 Chr.11: 123059308-123059308 on Build GRCh38
rs781053435 Chr.11: 123059409-123059409 on Build GRCh38
rs1033275003 Chr.11: 123059420-123059420 on Build GRCh38
rs780670105 Chr.11: 123059279-123059281 on Build GRCh38
rs764797484 Chr.11: 123059276-123059276 on Build GRCh38
rs757290841 Chr.11: 123059295-123059295 on Build GRCh38
rs145810402 Chr.11: 123059296-123059296 on Build GRCh38
rs748950534 Chr.11: 123059429-123059429 on Build GRCh38
rs376826236 Chr.11: 123059311-123059311 on Build GRCh38
rs746913030 Chr.11: 123059299-123059299 on Build GRCh38
rs554981599 Chr.11: 123059421-123059421 on Build GRCh38
rs111708230 Chr.11: 123059401-123059402 on Build GRCh38
rs575740356 Chr.11: 123059363-123059363 on Build GRCh38
rs200762609 Chr.11: 123059433-123059433 on Build GRCh38
rs1050909318 Chr.11: 123059329-123059330 on Build GRCh38
rs770829808 Chr.11: 123059394-123059396 on Build GRCh38
rs932006112 Chr.11: 123059325-123059325 on Build GRCh38
rs780901332 Chr.11: 123059418-123059421 on Build GRCh38
rs768454666 Chr.11: 123059264-123059264 on Build GRCh38
rs376026739 Chr.11: 123059277-123059277 on Build GRCh38
rs1020468229 Chr.11: 123059227-123059227 on Build GRCh38
rs752066770 Chr.11: 123059281-123059281 on Build GRCh38
rs111548556 Chr.11: 123059333-123059333 on Build GRCh38
rs990942244 Chr.11: 123059284-123059284 on Build GRCh38
rs775959865 Chr.11: 123059282-123059291 on Build GRCh38
rs760055366 Chr.11: 123059373-123059373 on Build GRCh38
rs371586742 Chr.11: 123059267-123059267 on Build GRCh38
rs755777776 Chr.11: 123059421-123059422 on Build GRCh38
rs746921647 Chr.11: 123059289-123059289 on Build GRCh38
rs762934951 Chr.11: 123059307-123059308 on Build GRCh38
rs778457298 Chr.11: 123059432-123059433 on Build GRCh38
rs751521507 Chr.11: 123059405-123059405 on Build GRCh38
rs899124471 Chr.11: 123059322-123059323 on Build GRCh38
rs202196042 Chr.11: 123059402-123059402 on Build GRCh38
rs552299685 Chr.11: 123059307-123059307 on Build GRCh38
rs375764038 Chr.11: 123059436-123059436 on Build GRCh38
rs531182299 Chr.11: 123059233-123059233 on Build GRCh38
rs1037706057 Chr.11: 123059321-123059321 on Build GRCh38
rs767902896 Chr.11: 123059285-123059285 on Build GRCh38
rs779642961 Chr.11: 123059385-123059385 on Build GRCh38
rs201862006 Chr.11: 123059398-123059399 on Build GRCh38
rs76152311 Chr.11: 123059400-123059401 on Build GRCh38
rs755322295 Chr.11: 123059282-123059282 on Build GRCh38
rs774771282 Chr.11: 123059372-123059372 on Build GRCh38
rs539172644 Chr.11: 123059334-123059334 on Build GRCh38
rs754933065 Chr.11: 123059407-123059407 on Build GRCh38
rs368933146 Chr.11: 123059265-123059265 on Build GRCh38
rs761063852 Chr.11: 123059391-123059391 on Build GRCh38
rs369832141 Chr.11: 123059312-123059312 on Build GRCh38
rs781040317 Chr.11: 123059306-123059306 on Build GRCh38
rs747945694 Chr.11: 123059413-123059413 on Build GRCh38
rs754399330 Chr.11: 123059414-123059415 on Build GRCh38
rs749047481 Chr.11: 123059433-123059436 on Build GRCh38
rs200246365 Chr.11: 123059399-123059400 on Build GRCh38
rs890561575 Chr.11: 123059337-123059337 on Build GRCh38
rs566723570 Chr.11: 123059292-123059292 on Build GRCh38
rs751114183 Chr.11: 123059402-123059405 on Build GRCh38
rs536635310 Chr.11: 123059394-123059394 on Build GRCh38
rs548113649 Chr.11: 123059279-123059279 on Build GRCh38
rs759659690 Chr.11: 123059263-123059264 on Build GRCh38
rs940623729 Chr.11: 123059313-123059313 on Build GRCh38
rs140240298 Chr.11: 123059419-123059420 on Build GRCh38
rs753006080 Chr.11: 123059286-123059286 on Build GRCh38
rs753097981 Chr.11: 123059387-123059387 on Build GRCh38
rs772986398 Chr.11: 123059294-123059295 on Build GRCh38
rs770390592 Chr.11: 123059432-123059432 on Build GRCh38

View Full Product Details