Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169509691-169509964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL41
Entrez Gene ID
10324
Gene Name
kelch like family member 41
Gene Aliases
KBTBD10, Krp1, SARCOSIN
UniGene
Hs.50550
Gene Chromosome Location
Chr.2: 169509702-169526262 on Build GRCh38
Transcripts
NM_006063.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6466455 c.8C>T p.S3F Chr.2: 169509786-169509786 on Build GRCh38
3569812 c.96C>T p.F32F Chr.2: 169509874-169509874 on Build GRCh38
3569811 c.37C>T p.L13F Chr.2: 169509815-169509815 on Build GRCh38
3315178 c.134C>T p.P45L Chr.2: 169509912-169509912 on Build GRCh38
1211363 c.175C>T p.R59C Chr.2: 169509953-169509953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs28730867 Chr.2: 169509786-169509786 on Build GRCh38
rs758830210 Chr.2: 169509763-169509763 on Build GRCh38
rs908739388 Chr.2: 169509722-169509722 on Build GRCh38
rs747281907 Chr.2: 169509773-169509773 on Build GRCh38
rs758522321 Chr.2: 169509842-169509842 on Build GRCh38
rs780361528 Chr.2: 169509843-169509843 on Build GRCh38
rs747822404 Chr.2: 169509792-169509792 on Build GRCh38
rs759362470 Chr.2: 169509804-169509804 on Build GRCh38
rs754142514 Chr.2: 169509739-169509739 on Build GRCh38
rs748552834 Chr.2: 169509859-169509859 on Build GRCh38
rs764344037 Chr.2: 169509734-169509734 on Build GRCh38
rs772566496 Chr.2: 169509796-169509796 on Build GRCh38
rs750461573 Chr.2: 169509840-169509840 on Build GRCh38
rs909848963 Chr.2: 169509727-169509727 on Build GRCh38
rs370341340 Chr.2: 169509897-169509897 on Build GRCh38
rs757361172 Chr.2: 169509743-169509743 on Build GRCh38
rs376586719 Chr.2: 169509851-169509851 on Build GRCh38
rs770503939 Chr.2: 169509920-169509920 on Build GRCh38
rs201785219 Chr.2: 169509791-169509791 on Build GRCh38
rs144048872 Chr.2: 169509793-169509793 on Build GRCh38
rs770715631 Chr.2: 169509801-169509801 on Build GRCh38
rs781233657 Chr.2: 169509913-169509913 on Build GRCh38
rs780672614 Chr.2: 169509769-169509769 on Build GRCh38
rs748678126 Chr.2: 169509903-169509903 on Build GRCh38
rs755289181 Chr.2: 169509856-169509856 on Build GRCh38
rs759387610 Chr.2: 169509729-169509729 on Build GRCh38
rs748332293 Chr.2: 169509726-169509726 on Build GRCh38
rs777495507 Chr.2: 169509901-169509901 on Build GRCh38
rs778040771 Chr.2: 169509902-169509903 on Build GRCh38
rs752585175 Chr.2: 169509828-169509828 on Build GRCh38
rs751918277 Chr.2: 169509780-169509780 on Build GRCh38
rs779110067 Chr.2: 169509751-169509751 on Build GRCh38
rs186952885 Chr.2: 169509754-169509754 on Build GRCh38
rs527886468 Chr.2: 169509813-169509813 on Build GRCh38
rs781712235 Chr.2: 169509789-169509789 on Build GRCh38
rs769016106 Chr.2: 169509776-169509776 on Build GRCh38
rs750552230 Chr.2: 169509753-169509753 on Build GRCh38
rs774105660 Chr.2: 169509933-169509933 on Build GRCh38
rs762581295 Chr.2: 169509797-169509797 on Build GRCh38
rs748591565 Chr.2: 169509880-169509880 on Build GRCh38
rs904435175 Chr.2: 169509864-169509864 on Build GRCh38
rs762092269 Chr.2: 169509830-169509830 on Build GRCh38
rs765617286 Chr.2: 169509839-169509839 on Build GRCh38

View Full Product Details