Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4932485-4932748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GP1BA
Entrez Gene ID
2811
Gene Name
glycoprotein Ib platelet alpha subunit
Gene Aliases
BDPLT1, BDPLT3, BSS, CD42B, CD42b-alpha, DBPLT3, GP1B, GPIbA, GPIbalpha, VWDP
UniGene
Hs.1472
Gene Chromosome Location
Chr.17: 4932275-4935030 on Build GRCh38
Transcripts
NM_000173.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4067581 c.63G>T p.E21D Chr.17: 4932667-4932667 on Build GRCh38
6147325 c.108G>A p.R36R Chr.17: 4932712-4932712 on Build GRCh38
4067580 c.40T>G p.L14V Chr.17: 4932644-4932644 on Build GRCh38
5430342 c.105G>A p.K35K Chr.17: 4932709-4932709 on Build GRCh38
4415069 c.51C>G p.H17Q Chr.17: 4932655-4932655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778779735 Chr.17: 4932575-4932575 on Build GRCh38
rs751416243 Chr.17: 4932671-4932671 on Build GRCh38
rs767711763 Chr.17: 4932561-4932561 on Build GRCh38
rs780863443 Chr.17: 4932681-4932681 on Build GRCh38
rs746692809 Chr.17: 4932584-4932584 on Build GRCh38
rs759849279 Chr.17: 4932636-4932636 on Build GRCh38
rs775619901 Chr.17: 4932724-4932724 on Build GRCh38
rs372373000 Chr.17: 4932641-4932641 on Build GRCh38
rs774650985 Chr.17: 4932634-4932634 on Build GRCh38
rs952083624 Chr.17: 4932649-4932649 on Build GRCh38
rs753658079 Chr.17: 4932569-4932569 on Build GRCh38
rs576387753 Chr.17: 4932513-4932513 on Build GRCh38
rs999674592 Chr.17: 4932550-4932550 on Build GRCh38
rs772389629 Chr.17: 4932720-4932720 on Build GRCh38
rs781032554 Chr.17: 4932611-4932613 on Build GRCh38
rs111292798 Chr.17: 4932710-4932710 on Build GRCh38
rs183989314 Chr.17: 4932577-4932577 on Build GRCh38
rs752489274 Chr.17: 4932717-4932717 on Build GRCh38
rs81663 Chr.17: 4932557-4932557 on Build GRCh38
rs773645699 Chr.17: 4932611-4932611 on Build GRCh38
rs758148266 Chr.17: 4932656-4932656 on Build GRCh38
rs755156892 Chr.17: 4932571-4932572 on Build GRCh38
rs778306339 Chr.17: 4932709-4932709 on Build GRCh38
rs974535671 Chr.17: 4932640-4932640 on Build GRCh38
rs866570250 Chr.17: 4932548-4932548 on Build GRCh38
rs751721207 Chr.17: 4932566-4932567 on Build GRCh38
rs911283855 Chr.17: 4932542-4932542 on Build GRCh38
rs958143327 Chr.17: 4932574-4932574 on Build GRCh38
rs201827537 Chr.17: 4932696-4932696 on Build GRCh38
rs554661306 Chr.17: 4932544-4932544 on Build GRCh38
rs770303624 Chr.17: 4932603-4932603 on Build GRCh38
rs992419285 Chr.17: 4932704-4932704 on Build GRCh38
rs760041588 Chr.17: 4932718-4932718 on Build GRCh38
rs376091002 Chr.17: 4932578-4932578 on Build GRCh38
rs544642889 Chr.17: 4932528-4932528 on Build GRCh38
rs2243093 Chr.17: 4932600-4932600 on Build GRCh38
rs774841072 Chr.17: 4932716-4932716 on Build GRCh38
rs768098161 Chr.17: 4932585-4932585 on Build GRCh38
rs754622081 Chr.17: 4932672-4932672 on Build GRCh38
rs747673131 Chr.17: 4932694-4932694 on Build GRCh38
rs767906261 Chr.17: 4932638-4932638 on Build GRCh38
rs965941179 Chr.17: 4932523-4932523 on Build GRCh38
rs763331024 Chr.17: 4932624-4932624 on Build GRCh38
rs771456681 Chr.17: 4932631-4932631 on Build GRCh38
rs765198146 Chr.17: 4932655-4932655 on Build GRCh38
rs757202817 Chr.17: 4932571-4932571 on Build GRCh38
rs560610176 Chr.17: 4932591-4932591 on Build GRCh38
rs779826628 Chr.17: 4932669-4932669 on Build GRCh38
rs760705175 Chr.17: 4932653-4932653 on Build GRCh38
rs771366895 Chr.17: 4932711-4932711 on Build GRCh38

View Full Product Details