Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108553571-108553806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1524
Entrez Gene ID
57650
Gene Name
KIAA1524
Gene Aliases
CIP2A, p90
UniGene
Hs.591308
Gene Chromosome Location
Chr.3: 108545752-108589644 on Build GRCh38
Transcripts
NM_020890.2 XM_006713716.3 XM_006713717.2 XM_011513056.1 XM_011513057.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5669136 c.2325-2A>G p.? Chr.3: 108553732-108553732 on Build GRCh38
4695704 c.2328T>G p.S776R Chr.3: 108553727-108553727 on Build GRCh38
7180852 c.2379G>A p.Q793Q Chr.3: 108553676-108553676 on Build GRCh38
6327020 c.2407+1G>A p.? Chr.3: 108553647-108553647 on Build GRCh38
3585003 c.2373G>A p.Q791Q Chr.3: 108553682-108553682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776083093 Chr.3: 108553615-108553615 on Build GRCh38
rs781589591 Chr.3: 108553718-108553718 on Build GRCh38
rs770155578 Chr.3: 108553614-108553614 on Build GRCh38
rs776839111 Chr.3: 108553739-108553740 on Build GRCh38
rs760886708 Chr.3: 108553679-108553680 on Build GRCh38
rs762304054 Chr.3: 108553623-108553623 on Build GRCh38
rs1044534225 Chr.3: 108553619-108553619 on Build GRCh38
rs758982264 Chr.3: 108553751-108553751 on Build GRCh38
rs764362092 Chr.3: 108553694-108553695 on Build GRCh38
rs757516656 Chr.3: 108553714-108553714 on Build GRCh38
rs766170273 Chr.3: 108553653-108553653 on Build GRCh38
rs771405897 Chr.3: 108553608-108553608 on Build GRCh38
rs766844883 Chr.3: 108553751-108553753 on Build GRCh38
rs200847374 Chr.3: 108553750-108553750 on Build GRCh38
rs750143113 Chr.3: 108553646-108553646 on Build GRCh38
rs746279061 Chr.3: 108553613-108553613 on Build GRCh38
rs751806817 Chr.3: 108553704-108553704 on Build GRCh38
rs767267426 Chr.3: 108553639-108553639 on Build GRCh38
rs752027191 Chr.3: 108553755-108553755 on Build GRCh38
rs756510345 Chr.3: 108553761-108553761 on Build GRCh38
rs184353560 Chr.3: 108553655-108553655 on Build GRCh38
rs749272185 Chr.3: 108553599-108553602 on Build GRCh38
rs761504119 Chr.3: 108553638-108553638 on Build GRCh38
rs750870941 Chr.3: 108553742-108553742 on Build GRCh38
rs770611748 Chr.3: 108553609-108553612 on Build GRCh38
rs1042337481 Chr.3: 108553611-108553611 on Build GRCh38
rs907365279 Chr.3: 108553640-108553640 on Build GRCh38
rs755406935 Chr.3: 108553756-108553757 on Build GRCh38
rs774173977 Chr.3: 108553634-108553636 on Build GRCh38
rs780420756 Chr.3: 108553773-108553773 on Build GRCh38
rs552946430 Chr.3: 108553648-108553648 on Build GRCh38
rs72470117 Chr.3: 108553741-108553742 on Build GRCh38
rs201134301 Chr.3: 108553636-108553636 on Build GRCh38
rs764323862 Chr.3: 108553703-108553703 on Build GRCh38
rs758543898 Chr.3: 108553701-108553701 on Build GRCh38
rs768132930 Chr.3: 108553775-108553775 on Build GRCh38
rs746164404 Chr.3: 108553742-108553742 on Build GRCh38
rs548589044 Chr.3: 108553774-108553774 on Build GRCh38
rs201915655 Chr.3: 108553631-108553631 on Build GRCh38
rs781720319 Chr.3: 108553609-108553609 on Build GRCh38
rs753813692 Chr.3: 108553659-108553659 on Build GRCh38

View Full Product Details