Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122716222-122716485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCAR3
Entrez Gene ID
8843
Gene Name
hydroxycarboxylic acid receptor 3
Gene Aliases
GPR109B, HCA3, HM74, PUMAG, Puma-g
UniGene
Hs.458425
Gene Chromosome Location
Chr.12: 122714756-122716892 on Build GRCh38
Transcripts
NM_006018.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6846304 c.289G>A p.D97N Chr.12: 122716449-122716449 on Build GRCh38
5980408 c.424T>C p.W142R Chr.12: 122716314-122716314 on Build GRCh38
6677837 c.457G>A p.G153S Chr.12: 122716281-122716281 on Build GRCh38
6194688 c.332G>T p.R111L Chr.12: 122716406-122716406 on Build GRCh38
2225241 c.382C>T p.R128W Chr.12: 122716356-122716356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3825146 Chr.12: 122716465-122716465 on Build GRCh38
rs564762924 Chr.12: 122716406-122716406 on Build GRCh38
rs367957885 Chr.12: 122716272-122716272 on Build GRCh38
rs759856375 Chr.12: 122716408-122716408 on Build GRCh38
rs1696352 Chr.12: 122716390-122716390 on Build GRCh38
rs769456602 Chr.12: 122716300-122716300 on Build GRCh38
rs146381939 Chr.12: 122716442-122716442 on Build GRCh38
rs761798559 Chr.12: 122716294-122716295 on Build GRCh38
rs374808951 Chr.12: 122716264-122716264 on Build GRCh38
rs373400007 Chr.12: 122716414-122716414 on Build GRCh38
rs753573884 Chr.12: 122716433-122716433 on Build GRCh38
rs371876470 Chr.12: 122716356-122716356 on Build GRCh38
rs755588864 Chr.12: 122716428-122716428 on Build GRCh38
rs768768321 Chr.12: 122716309-122716309 on Build GRCh38
rs182089499 Chr.12: 122716315-122716315 on Build GRCh38
rs531947529 Chr.12: 122716437-122716437 on Build GRCh38
rs202161035 Chr.12: 122716314-122716314 on Build GRCh38
rs375685539 Chr.12: 122716334-122716334 on Build GRCh38
rs924289673 Chr.12: 122716431-122716431 on Build GRCh38
rs924501811 Chr.12: 122716256-122716256 on Build GRCh38
rs777701516 Chr.12: 122716451-122716451 on Build GRCh38
rs529768166 Chr.12: 122716342-122716342 on Build GRCh38
rs1040966998 Chr.12: 122716440-122716440 on Build GRCh38
rs201704809 Chr.12: 122716436-122716436 on Build GRCh38
rs773257135 Chr.12: 122716316-122716316 on Build GRCh38
rs753183372 Chr.12: 122716419-122716419 on Build GRCh38
rs557508973 Chr.12: 122716374-122716374 on Build GRCh38
rs200377324 Chr.12: 122716430-122716430 on Build GRCh38
rs977516577 Chr.12: 122716407-122716407 on Build GRCh38
rs560627929 Chr.12: 122716373-122716373 on Build GRCh38
rs761003966 Chr.12: 122716320-122716320 on Build GRCh38
rs780732393 Chr.12: 122716355-122716355 on Build GRCh38
rs772254116 Chr.12: 122716382-122716382 on Build GRCh38
rs375376257 Chr.12: 122716393-122716393 on Build GRCh38
rs769293992 Chr.12: 122716353-122716353 on Build GRCh38
rs763493478 Chr.12: 122716423-122716423 on Build GRCh38
rs775398822 Chr.12: 122716301-122716301 on Build GRCh38
rs541577169 Chr.12: 122716360-122716360 on Build GRCh38
rs750011531 Chr.12: 122716425-122716425 on Build GRCh38
rs948723144 Chr.12: 122716362-122716362 on Build GRCh38
rs116605431 Chr.12: 122716303-122716303 on Build GRCh38
rs771127331 Chr.12: 122716404-122716404 on Build GRCh38
rs3825145 Chr.12: 122716454-122716454 on Build GRCh38
rs368480408 Chr.12: 122716336-122716336 on Build GRCh38
rs745559851 Chr.12: 122716276-122716276 on Build GRCh38
rs755549091 Chr.12: 122716243-122716243 on Build GRCh38
rs748118129 Chr.12: 122716381-122716381 on Build GRCh38
rs756709608 Chr.12: 122716353-122716353 on Build GRCh38
rs1041307119 Chr.12: 122716270-122716270 on Build GRCh38
rs376548365 Chr.12: 122716435-122716435 on Build GRCh38
rs766622000 Chr.12: 122716323-122716323 on Build GRCh38
rs370219885 Chr.12: 122716292-122716292 on Build GRCh38
rs772202813 Chr.12: 122716439-122716439 on Build GRCh38
rs376177871 Chr.12: 122716372-122716372 on Build GRCh38
rs959488319 Chr.12: 122716357-122716357 on Build GRCh38
rs996923561 Chr.12: 122716291-122716291 on Build GRCh38

View Full Product Details