Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116886407-116886660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO4
Entrez Gene ID
387712
Gene Name
enolase family member 4
Gene Aliases
C10orf134
UniGene
Hs.693248
Gene Chromosome Location
Chr.10: 116848230-116911803 on Build GRCh38
Transcripts
XM_006717835.3 NM_001127211.2 NM_001258298.1 NM_018330.6 XM_006717931.3 XM_017016462.1

Gene Symbol
SHTN1
Entrez Gene ID
57698
Gene Name
shootin 1
Gene Aliases
KIAA1598
UniGene
Hs.501140
Gene Chromosome Location
Chr.10: 116883377-117126586 on Build GRCh38
Transcripts
XM_006717835.3 NM_001127211.2 NM_001258298.1 NM_018330.6 XM_006717931.3 XM_017016462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5789446 c.1729G>A p.E577K Chr.10: 116886511-116886511 on Build GRCh38
465314 c.1828G>A p.E610K Chr.10: 116886412-116886412 on Build GRCh38
5131062 c.1704_1705insA p.Y569fs*3 Chr.10: 116886535-116886536 on Build GRCh38
7425877 c.1769C>T p.P590L Chr.10: 116886471-116886471 on Build GRCh38
4997271 c.1744T>G p.L582V Chr.10: 116886496-116886496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751085650 Chr.10: 116886454-116886454 on Build GRCh38
rs377600413 Chr.10: 116886512-116886512 on Build GRCh38
rs767108544 Chr.10: 116886591-116886591 on Build GRCh38
rs754109130 Chr.10: 116886490-116886490 on Build GRCh38
rs775337559 Chr.10: 116886430-116886430 on Build GRCh38
rs778054942 Chr.10: 116886604-116886605 on Build GRCh38
rs2531685 Chr.10: 116886618-116886618 on Build GRCh38
rs551803965 Chr.10: 116886552-116886552 on Build GRCh38
rs763899193 Chr.10: 116886448-116886448 on Build GRCh38
rs373921504 Chr.10: 116886579-116886579 on Build GRCh38
rs775388229 Chr.10: 116886531-116886531 on Build GRCh38
rs890364912 Chr.10: 116886566-116886566 on Build GRCh38
rs748204859 Chr.10: 116886587-116886588 on Build GRCh38
rs750098990 Chr.10: 116886472-116886472 on Build GRCh38
rs755501920 Chr.10: 116886583-116886584 on Build GRCh38
rs199659749 Chr.10: 116886609-116886609 on Build GRCh38
rs866759482 Chr.10: 116886456-116886456 on Build GRCh38
rs778884339 Chr.10: 116886588-116886588 on Build GRCh38
rs149273896 Chr.10: 116886596-116886597 on Build GRCh38
rs572392240 Chr.10: 116886516-116886517 on Build GRCh38
rs1043130751 Chr.10: 116886439-116886439 on Build GRCh38
rs761485147 Chr.10: 116886581-116886581 on Build GRCh38
rs368223618 Chr.10: 116886522-116886522 on Build GRCh38
rs755510375 Chr.10: 116886501-116886501 on Build GRCh38
rs756589614 Chr.10: 116886477-116886477 on Build GRCh38
rs761174075 Chr.10: 116886466-116886466 on Build GRCh38
rs768652780 Chr.10: 116886571-116886571 on Build GRCh38
rs767339763 Chr.10: 116886470-116886470 on Build GRCh38
rs773050256 Chr.10: 116886599-116886599 on Build GRCh38
rs984007640 Chr.10: 116886467-116886467 on Build GRCh38
rs952623389 Chr.10: 116886449-116886449 on Build GRCh38
rs780585711 Chr.10: 116886483-116886483 on Build GRCh38
rs772133798 Chr.10: 116886517-116886517 on Build GRCh38
rs200994601 Chr.10: 116886526-116886526 on Build GRCh38
rs779225990 Chr.10: 116886511-116886511 on Build GRCh38
rs111941372 Chr.10: 116886527-116886527 on Build GRCh38

View Full Product Details