Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99397494-99397737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOT1
Entrez Gene ID
2805
Gene Name
glutamic-oxaloacetic transaminase 1
Gene Aliases
AST1, ASTQTL1, GIG18, cAspAT, cCAT
UniGene
Hs.500756
Gene Chromosome Location
Chr.10: 99396870-99430773 on Build GRCh38
Transcripts
NM_002079.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
281434 c.1160G>A p.R387Q Chr.10: 99397629-99397629 on Build GRCh38
6460822 c.1167C>T p.N389N Chr.10: 99397622-99397622 on Build GRCh38
4010922 c.1134G>A p.K378K Chr.10: 99397655-99397655 on Build GRCh38
4010921 c.1222G>A p.A408T Chr.10: 99397567-99397567 on Build GRCh38
7215205 c.1108C>G p.Q370E Chr.10: 99397681-99397681 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763011514 Chr.10: 99397671-99397671 on Build GRCh38
rs769642084 Chr.10: 99397621-99397621 on Build GRCh38
rs762024546 Chr.10: 99397712-99397712 on Build GRCh38
rs772362314 Chr.10: 99397534-99397534 on Build GRCh38
rs571090139 Chr.10: 99397589-99397589 on Build GRCh38
rs770543768 Chr.10: 99397565-99397565 on Build GRCh38
rs776110451 Chr.10: 99397709-99397710 on Build GRCh38
rs371927397 Chr.10: 99397588-99397588 on Build GRCh38
rs767829544 Chr.10: 99397529-99397529 on Build GRCh38
rs150817762 Chr.10: 99397673-99397673 on Build GRCh38
rs200910674 Chr.10: 99397579-99397579 on Build GRCh38
rs757866070 Chr.10: 99397541-99397541 on Build GRCh38
rs376753781 Chr.10: 99397519-99397519 on Build GRCh38
rs1030506758 Chr.10: 99397557-99397557 on Build GRCh38
rs551762102 Chr.10: 99397550-99397550 on Build GRCh38
rs76850691 Chr.10: 99397681-99397681 on Build GRCh38
rs868066178 Chr.10: 99397655-99397655 on Build GRCh38
rs775222620 Chr.10: 99397622-99397622 on Build GRCh38
rs142360515 Chr.10: 99397542-99397542 on Build GRCh38
rs368527371 Chr.10: 99397533-99397533 on Build GRCh38
rs780967129 Chr.10: 99397583-99397583 on Build GRCh38
rs749913156 Chr.10: 99397622-99397624 on Build GRCh38
rs1021532307 Chr.10: 99397677-99397677 on Build GRCh38
rs559712292 Chr.10: 99397536-99397536 on Build GRCh38
rs747300745 Chr.10: 99397592-99397592 on Build GRCh38
rs547920235 Chr.10: 99397535-99397535 on Build GRCh38

View Full Product Details