Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 127595109-127595368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCN3
Entrez Gene ID
29999
Gene Name
fascin actin-bundling protein 3
Gene Aliases
-
UniGene
Hs.712692
Gene Chromosome Location
Chr.7: 127593635-127601797 on Build GRCh38
Transcripts
NM_020369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3632952 c.150G>A p.W50* Chr.7: 127595312-127595312 on Build GRCh38
3632953 c.185C>T p.A62V Chr.7: 127595347-127595347 on Build GRCh38
123263 c.187G>A p.V63M Chr.7: 127595349-127595349 on Build GRCh38
5660145 c.154A>T p.I52F Chr.7: 127595316-127595316 on Build GRCh38
744674 c.145-1G>A p.? Chr.7: 127595306-127595306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763699964 Chr.7: 127595336-127595336 on Build GRCh38
rs774950391 Chr.7: 127595250-127595250 on Build GRCh38
rs1039497060 Chr.7: 127595254-127595254 on Build GRCh38
rs987238057 Chr.7: 127595196-127595196 on Build GRCh38
rs1052104705 Chr.7: 127595212-127595212 on Build GRCh38
rs935083310 Chr.7: 127595211-127595211 on Build GRCh38
rs960292101 Chr.7: 127595328-127595328 on Build GRCh38
rs775073701 Chr.7: 127595332-127595332 on Build GRCh38
rs540448316 Chr.7: 127595258-127595258 on Build GRCh38
rs773652430 Chr.7: 127595313-127595313 on Build GRCh38
rs772390689 Chr.7: 127595300-127595300 on Build GRCh38
rs1048876688 Chr.7: 127595307-127595307 on Build GRCh38
rs768778971 Chr.7: 127595267-127595267 on Build GRCh38
rs779337876 Chr.7: 127595269-127595269 on Build GRCh38
rs147063403 Chr.7: 127595174-127595174 on Build GRCh38
rs780174647 Chr.7: 127595259-127595259 on Build GRCh38
rs924969960 Chr.7: 127595158-127595158 on Build GRCh38
rs150511464 Chr.7: 127595331-127595331 on Build GRCh38
rs748661094 Chr.7: 127595270-127595270 on Build GRCh38
rs760995892 Chr.7: 127595322-127595322 on Build GRCh38
rs368213335 Chr.7: 127595266-127595266 on Build GRCh38
rs138368376 Chr.7: 127595349-127595349 on Build GRCh38
rs1018824706 Chr.7: 127595225-127595225 on Build GRCh38
rs762540012 Chr.7: 127595335-127595335 on Build GRCh38
rs768621705 Chr.7: 127595282-127595283 on Build GRCh38
rs139948688 Chr.7: 127595348-127595348 on Build GRCh38
rs550698572 Chr.7: 127595249-127595249 on Build GRCh38

View Full Product Details