Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138610911-138611178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAIM
Entrez Gene ID
55179
Gene Name
Fas apoptotic inhibitory molecule
Gene Aliases
FAIM1
UniGene
Hs.173438
Gene Chromosome Location
Chr.3: 138608163-138633376 on Build GRCh38
Transcripts
NM_001033030.1 NM_001033031.1 NM_001033032.1 NM_018147.3 XM_011512950.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7107565 c.63G>A p.P21P Chr.3: 138611020-138611020 on Build GRCh38
5059357 c.71C>T p.A24V Chr.3: 138611028-138611028 on Build GRCh38
6468487 c.36G>A p.L12L Chr.3: 138610993-138610993 on Build GRCh38
4461283 c.11C>T p.P4L Chr.3: 138610968-138610968 on Build GRCh38
6242591 c.33G>A p.L11L Chr.3: 138610990-138610990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367804345 Chr.3: 138610961-138610961 on Build GRCh38
rs548137847 Chr.3: 138611137-138611137 on Build GRCh38
rs762543545 Chr.3: 138611062-138611062 on Build GRCh38
rs139741354 Chr.3: 138611010-138611010 on Build GRCh38
rs767213910 Chr.3: 138611076-138611076 on Build GRCh38
rs375438065 Chr.3: 138611087-138611087 on Build GRCh38
rs776041850 Chr.3: 138611029-138611029 on Build GRCh38
rs370708515 Chr.3: 138611089-138611089 on Build GRCh38
rs771121254 Chr.3: 138610984-138610984 on Build GRCh38
rs140708934 Chr.3: 138611069-138611069 on Build GRCh38
rs973249683 Chr.3: 138611146-138611146 on Build GRCh38
rs811322 Chr.3: 138611020-138611020 on Build GRCh38
rs749535650 Chr.3: 138610964-138610964 on Build GRCh38
rs908635268 Chr.3: 138611123-138611123 on Build GRCh38
rs751187212 Chr.3: 138611042-138611042 on Build GRCh38
rs964851671 Chr.3: 138611022-138611022 on Build GRCh38
rs764737445 Chr.3: 138611049-138611049 on Build GRCh38
rs746197659 Chr.3: 138611019-138611019 on Build GRCh38
rs902905647 Chr.3: 138610966-138610966 on Build GRCh38
rs761154458 Chr.3: 138611037-138611037 on Build GRCh38
rs781237469 Chr.3: 138610955-138610955 on Build GRCh38
rs756330219 Chr.3: 138610960-138610960 on Build GRCh38
rs751328940 Chr.3: 138610941-138610941 on Build GRCh38
rs772639483 Chr.3: 138611058-138611058 on Build GRCh38
rs779713614 Chr.3: 138610937-138610937 on Build GRCh38
rs766006461 Chr.3: 138611065-138611065 on Build GRCh38
rs748065294 Chr.3: 138610958-138610958 on Build GRCh38
rs747985979 Chr.3: 138611023-138611024 on Build GRCh38
rs368013436 Chr.3: 138610953-138610953 on Build GRCh38
rs754840071 Chr.3: 138611072-138611072 on Build GRCh38

View Full Product Details