Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41726960-41727233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAP1
Entrez Gene ID
9001
Gene Name
huntingtin associated protein 1
Gene Aliases
HAP2, HIP5, HLP, hHLP1
UniGene
Hs.158300
Gene Chromosome Location
Chr.17: 41722639-41734646 on Build GRCh38
Transcripts
NM_001079870.1 NM_001079871.1 NM_177977.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5891510 c.1362G>A p.M454I Chr.17: 41727058-41727058 on Build GRCh38
3889671 c.1334G>A p.R445K Chr.17: 41727086-41727086 on Build GRCh38
6252135 c.1347C>A p.D449E Chr.17: 41727073-41727073 on Build GRCh38
7334999 c.1266C>T p.L422L Chr.17: 41727103-41727103 on Build GRCh38
4985792 c.1478C>T p.T493M Chr.17: 41727098-41727098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782318727 Chr.17: 41727188-41727188 on Build GRCh38
rs782457736 Chr.17: 41727094-41727094 on Build GRCh38
rs922423118 Chr.17: 41727176-41727176 on Build GRCh38
rs376195729 Chr.17: 41727135-41727135 on Build GRCh38
rs955217927 Chr.17: 41727180-41727180 on Build GRCh38
rs782231666 Chr.17: 41727072-41727072 on Build GRCh38
rs955843093 Chr.17: 41727168-41727168 on Build GRCh38
rs782496737 Chr.17: 41727155-41727155 on Build GRCh38
rs782546957 Chr.17: 41727119-41727119 on Build GRCh38
rs782004258 Chr.17: 41727102-41727102 on Build GRCh38
rs896728289 Chr.17: 41727117-41727117 on Build GRCh38
rs568933180 Chr.17: 41727165-41727165 on Build GRCh38
rs201427462 Chr.17: 41727134-41727134 on Build GRCh38
rs889752905 Chr.17: 41727022-41727022 on Build GRCh38
rs372575811 Chr.17: 41727027-41727027 on Build GRCh38
rs985317728 Chr.17: 41727194-41727194 on Build GRCh38
rs376650931 Chr.17: 41727111-41727111 on Build GRCh38
rs367805108 Chr.17: 41727164-41727164 on Build GRCh38
rs1049637139 Chr.17: 41727013-41727013 on Build GRCh38
rs782789933 Chr.17: 41727149-41727149 on Build GRCh38
rs372630337 Chr.17: 41727010-41727010 on Build GRCh38
rs782468634 Chr.17: 41727177-41727177 on Build GRCh38
rs782667844 Chr.17: 41727096-41727096 on Build GRCh38
rs377587501 Chr.17: 41727048-41727048 on Build GRCh38
rs200864908 Chr.17: 41727174-41727174 on Build GRCh38
rs782182576 Chr.17: 41727186-41727186 on Build GRCh38
rs781844556 Chr.17: 41727095-41727095 on Build GRCh38
rs782509605 Chr.17: 41727051-41727051 on Build GRCh38
rs782697385 Chr.17: 41727162-41727162 on Build GRCh38
rs782470877 Chr.17: 41727097-41727097 on Build GRCh38
rs369093232 Chr.17: 41727049-41727049 on Build GRCh38
rs782532528 Chr.17: 41727040-41727040 on Build GRCh38
rs782633188 Chr.17: 41727178-41727178 on Build GRCh38
rs187822559 Chr.17: 41727017-41727017 on Build GRCh38
rs4523977 Chr.17: 41727098-41727098 on Build GRCh38
rs782797847 Chr.17: 41727020-41727020 on Build GRCh38
rs143945817 Chr.17: 41727077-41727077 on Build GRCh38
rs199819567 Chr.17: 41727057-41727057 on Build GRCh38
rs782105086 Chr.17: 41727148-41727148 on Build GRCh38
rs782031193 Chr.17: 41727142-41727142 on Build GRCh38
rs7211719 Chr.17: 41727024-41727024 on Build GRCh38
rs782693741 Chr.17: 41727034-41727034 on Build GRCh38
rs781819059 Chr.17: 41727150-41727150 on Build GRCh38
rs782010018 Chr.17: 41727191-41727191 on Build GRCh38
rs528686279 Chr.17: 41727036-41727036 on Build GRCh38
rs782291160 Chr.17: 41727196-41727196 on Build GRCh38
rs782773222 Chr.17: 41727134-41727134 on Build GRCh38
rs374580379 Chr.17: 41727118-41727118 on Build GRCh38
rs969680701 Chr.17: 41727157-41727157 on Build GRCh38
rs558587462 Chr.17: 41727175-41727175 on Build GRCh38
rs941347237 Chr.17: 41727204-41727204 on Build GRCh38
rs933950033 Chr.17: 41726982-41726982 on Build GRCh38

View Full Product Details