Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 152963028-152963253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHDC1
Entrez Gene ID
85462
Gene Name
FH2 domain containing 1
Gene Aliases
-
UniGene
Hs.132629
Gene Chromosome Location
Chr.4: 152911570-152979696 on Build GRCh38
Transcripts
NM_033393.2 XM_005263319.3 XM_006714393.3 XM_011532388.1 XM_011532389.1 XM_017008787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5810670 c.1028A>T p.Q343L Chr.4: 152963129-152963129 on Build GRCh38
5865889 c.943G>A p.V315I Chr.4: 152963044-152963044 on Build GRCh38
7078048 c.934G>A p.G312S Chr.4: 152963035-152963035 on Build GRCh38
5865890 c.962C>T p.S321F Chr.4: 152963063-152963063 on Build GRCh38
4919848 c.982A>G p.T328A Chr.4: 152963083-152963083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111713832 Chr.4: 152963061-152963061 on Build GRCh38
rs916607252 Chr.4: 152963158-152963158 on Build GRCh38
rs1046709191 Chr.4: 152963181-152963181 on Build GRCh38
rs767171131 Chr.4: 152963113-152963113 on Build GRCh38
rs376527715 Chr.4: 152963182-152963182 on Build GRCh38
rs551020048 Chr.4: 152963215-152963215 on Build GRCh38
rs1034199652 Chr.4: 152963163-152963163 on Build GRCh38
rs749498428 Chr.4: 152963059-152963059 on Build GRCh38
rs773547122 Chr.4: 152963057-152963057 on Build GRCh38
rs375603714 Chr.4: 152963160-152963160 on Build GRCh38
rs774301989 Chr.4: 152963064-152963064 on Build GRCh38
rs892751106 Chr.4: 152963060-152963060 on Build GRCh38
rs776613833 Chr.4: 152963094-152963094 on Build GRCh38
rs373735764 Chr.4: 152963074-152963074 on Build GRCh38
rs990827212 Chr.4: 152963114-152963114 on Build GRCh38
rs368956300 Chr.4: 152963171-152963171 on Build GRCh38
rs935506032 Chr.4: 152963198-152963198 on Build GRCh38
rs762545186 Chr.4: 152963166-152963166 on Build GRCh38
rs370344776 Chr.4: 152963052-152963052 on Build GRCh38
rs963374040 Chr.4: 152963210-152963210 on Build GRCh38
rs768564454 Chr.4: 152963080-152963080 on Build GRCh38
rs765062035 Chr.4: 152963161-152963161 on Build GRCh38

View Full Product Details