Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119120284-119120511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM45A
Entrez Gene ID
404636
Gene Name
family with sequence similarity 45 member A
Gene Aliases
-
UniGene
Hs.434241
Gene Chromosome Location
Chr.10: 119104065-119137984 on Build GRCh38
Transcripts
NM_001303111.1 NM_001303112.1 NM_001303113.1 NM_207009.3 NR_130122.1 XM_006717849.3 XM_006717850.3 XM_006717851.3 XM_006717852.3 XM_017016261.1 XM_017016262.1 XM_017016263.1 XM_017016264.1 XM_017016265.1 XM_017016266.1 XM_017016267.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1739650 c.537A>G p.L179L Chr.10: 119120396-119120396 on Build GRCh38
6307692 c.500G>T p.G167V Chr.10: 119120359-119120359 on Build GRCh38
5620788 c.491C>T p.S164F Chr.10: 119120350-119120350 on Build GRCh38
6485893 c.514A>G p.I172V Chr.10: 119120373-119120373 on Build GRCh38
915339 c.545G>T p.R182I Chr.10: 119120404-119120404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005167159 Chr.10: 119120376-119120376 on Build GRCh38
rs552500096 Chr.10: 119120329-119120329 on Build GRCh38
rs773548339 Chr.10: 119120414-119120414 on Build GRCh38
rs752650472 Chr.10: 119120437-119120437 on Build GRCh38
rs938731704 Chr.10: 119120442-119120442 on Build GRCh38
rs565986270 Chr.10: 119120383-119120383 on Build GRCh38
rs554487974 Chr.10: 119120407-119120407 on Build GRCh38
rs764505405 Chr.10: 119120343-119120343 on Build GRCh38
rs189634789 Chr.10: 119120478-119120478 on Build GRCh38
rs777751681 Chr.10: 119120328-119120328 on Build GRCh38
rs146464623 Chr.10: 119120408-119120408 on Build GRCh38
rs112756319 Chr.10: 119120474-119120474 on Build GRCh38
rs369346358 Chr.10: 119120379-119120379 on Build GRCh38
rs760822569 Chr.10: 119120420-119120420 on Build GRCh38
rs181997301 Chr.10: 119120482-119120482 on Build GRCh38
rs780697204 Chr.10: 119120341-119120341 on Build GRCh38
rs752321728 Chr.10: 119120453-119120454 on Build GRCh38
rs372966495 Chr.10: 119120392-119120392 on Build GRCh38
rs555162586 Chr.10: 119120447-119120447 on Build GRCh38
rs780530031 Chr.10: 119120360-119120360 on Build GRCh38
rs144364814 Chr.10: 119120359-119120359 on Build GRCh38
rs778433153 Chr.10: 119120347-119120347 on Build GRCh38
rs756884347 Chr.10: 119120333-119120333 on Build GRCh38
rs749829928 Chr.10: 119120368-119120368 on Build GRCh38
rs766614927 Chr.10: 119120435-119120435 on Build GRCh38
rs769363889 Chr.10: 119120354-119120354 on Build GRCh38
rs754511890 Chr.10: 119120343-119120345 on Build GRCh38
rs963053497 Chr.10: 119120390-119120390 on Build GRCh38

View Full Product Details