Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92456268-92456527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDE
Entrez Gene ID
3416
Gene Name
insulin degrading enzyme
Gene Aliases
INSULYSIN
UniGene
Hs.500546
Gene Chromosome Location
Chr.10: 92451684-92574484 on Build GRCh38
Transcripts
NM_001165946.1 NM_001322793.1 NM_001322794.1 NM_001322795.1 NM_001322796.1 NM_001322797.1 NM_004969.3 NR_136399.1 XM_017016187.1 XM_017016188.1 XM_017016189.1 XM_017016190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3665954 c.2846C>G p.P949R Chr.10: 92456409-92456409 on Build GRCh38
5837454 c.2832G>T p.L944F Chr.10: 92456423-92456423 on Build GRCh38
921462 c.2861T>C p.V954A Chr.10: 92456394-92456394 on Build GRCh38
5904927 c.2846C>T p.P949L Chr.10: 92456409-92456409 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752286492 Chr.10: 92456310-92456310 on Build GRCh38
rs965763686 Chr.10: 92456358-92456358 on Build GRCh38
rs556005554 Chr.10: 92456318-92456318 on Build GRCh38
rs770605852 Chr.10: 92456364-92456364 on Build GRCh38
rs777078112 Chr.10: 92456395-92456395 on Build GRCh38
rs780296196 Chr.10: 92456374-92456374 on Build GRCh38
rs201935226 Chr.10: 92456304-92456304 on Build GRCh38
rs200095067 Chr.10: 92456448-92456448 on Build GRCh38
rs949313805 Chr.10: 92456488-92456488 on Build GRCh38
rs769078696 Chr.10: 92456390-92456390 on Build GRCh38
rs749017719 Chr.10: 92456343-92456343 on Build GRCh38
rs767583086 Chr.10: 92456478-92456478 on Build GRCh38
rs369371410 Chr.10: 92456407-92456407 on Build GRCh38
rs182056762 Chr.10: 92456462-92456462 on Build GRCh38
rs907662658 Chr.10: 92456355-92456355 on Build GRCh38
rs12356364 Chr.10: 92456416-92456416 on Build GRCh38
rs188796899 Chr.10: 92456327-92456327 on Build GRCh38
rs375559999 Chr.10: 92456389-92456389 on Build GRCh38
rs372148441 Chr.10: 92456319-92456319 on Build GRCh38
rs769903924 Chr.10: 92456429-92456429 on Build GRCh38
rs762289249 Chr.10: 92456406-92456406 on Build GRCh38
rs759334464 Chr.10: 92456476-92456476 on Build GRCh38
rs774175817 Chr.10: 92456335-92456335 on Build GRCh38
rs755635595 Chr.10: 92456314-92456314 on Build GRCh38
rs773274122 Chr.10: 92456435-92456435 on Build GRCh38
rs201758660 Chr.10: 92456433-92456433 on Build GRCh38
rs781209506 Chr.10: 92456309-92456309 on Build GRCh38
rs774017679 Chr.10: 92456473-92456473 on Build GRCh38

View Full Product Details