Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124919692-124919961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEPACAM
Entrez Gene ID
220296
Gene Name
hepatic and glial cell adhesion molecule
Gene Aliases
GlialCAM, MLC2A, MLC2B
UniGene
Hs.745294
Gene Chromosome Location
Chr.11: 124919244-124936412 on Build GRCh38
Transcripts
NM_152722.4 XM_005271449.2 XM_017017361.1 NM_001037558.2

Gene Symbol
HEPN1
Entrez Gene ID
641654
Gene Name
hepatocellular carcinoma, down-regulated 1
Gene Aliases
-
UniGene
Hs.159863
Gene Chromosome Location
Chr.11: 124919250-124920677 on Build GRCh38
Transcripts
NM_152722.4 XM_005271449.2 XM_017017361.1 NM_001037558.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
924461 c.172G>A p.D58N Chr.11: 124919922-124919922 on Build GRCh38
924460 c.79G>A p.G27R Chr.11: 124919829-124919829 on Build GRCh38
1317360 c.182A>T p.H61L Chr.11: 124919932-124919932 on Build GRCh38
1321812 c.198C>T p.F66F Chr.11: 124919948-124919948 on Build GRCh38
1676557 c.206G>A p.R69Q Chr.11: 124919956-124919956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773210098 Chr.11: 124919784-124919784 on Build GRCh38
rs758244596 Chr.11: 124919727-124919727 on Build GRCh38
rs561678478 Chr.11: 124919915-124919915 on Build GRCh38
rs749383944 Chr.11: 124919855-124919855 on Build GRCh38
rs572317793 Chr.11: 124919805-124919805 on Build GRCh38
rs938319629 Chr.11: 124919910-124919910 on Build GRCh38
rs764567897 Chr.11: 124919885-124919885 on Build GRCh38
rs758177602 Chr.11: 124919938-124919938 on Build GRCh38
rs778846572 Chr.11: 124919739-124919739 on Build GRCh38
rs780074085 Chr.11: 124919750-124919750 on Build GRCh38
rs368280630 Chr.11: 124919792-124919792 on Build GRCh38
rs1037519522 Chr.11: 124919935-124919935 on Build GRCh38
rs745717656 Chr.11: 124919740-124919740 on Build GRCh38
rs539370875 Chr.11: 124919806-124919806 on Build GRCh38
rs756900952 Chr.11: 124919736-124919736 on Build GRCh38
rs772008421 Chr.11: 124919742-124919742 on Build GRCh38
rs774682077 Chr.11: 124919795-124919795 on Build GRCh38
rs764875323 Chr.11: 124919930-124919930 on Build GRCh38
rs201393780 Chr.11: 124919904-124919904 on Build GRCh38
rs777210646 Chr.11: 124919896-124919896 on Build GRCh38
rs963356487 Chr.11: 124919895-124919895 on Build GRCh38
rs914743928 Chr.11: 124919856-124919856 on Build GRCh38
rs886222939 Chr.11: 124919919-124919919 on Build GRCh38
rs987309911 Chr.11: 124919741-124919741 on Build GRCh38
rs761240901 Chr.11: 124919879-124919879 on Build GRCh38
rs780179144 Chr.11: 124919793-124919793 on Build GRCh38
rs202007726 Chr.11: 124919942-124919942 on Build GRCh38
rs773166911 Chr.11: 124919756-124919756 on Build GRCh38
rs777901233 Chr.11: 124919850-124919850 on Build GRCh38
rs3802904 Chr.11: 124919859-124919859 on Build GRCh38
rs763029230 Chr.11: 124919760-124919761 on Build GRCh38
rs761108320 Chr.11: 124919807-124919807 on Build GRCh38
rs200402085 Chr.11: 124919820-124919820 on Build GRCh38
rs759835890 Chr.11: 124919798-124919798 on Build GRCh38
rs370843348 Chr.11: 124919822-124919822 on Build GRCh38
rs750107338 Chr.11: 124919833-124919833 on Build GRCh38
rs527377754 Chr.11: 124919847-124919849 on Build GRCh38
rs775766828 Chr.11: 124919872-124919872 on Build GRCh38
rs1016047270 Chr.11: 124919873-124919873 on Build GRCh38
rs763629377 Chr.11: 124919815-124919815 on Build GRCh38
rs761163524 Chr.11: 124919928-124919928 on Build GRCh38
rs768512422 Chr.11: 124919848-124919848 on Build GRCh38
rs768666007 Chr.11: 124919920-124919920 on Build GRCh38
rs201002336 Chr.11: 124919763-124919763 on Build GRCh38
rs1000899669 Chr.11: 124919744-124919746 on Build GRCh38
rs78859654 Chr.11: 124919932-124919932 on Build GRCh38
rs903865485 Chr.11: 124919726-124919726 on Build GRCh38
rs772373154 Chr.11: 124919861-124919861 on Build GRCh38
rs201178398 Chr.11: 124919860-124919860 on Build GRCh38
rs753356212 Chr.11: 124919818-124919818 on Build GRCh38
rs146650696 Chr.11: 124919838-124919838 on Build GRCh38
rs375152348 Chr.11: 124919760-124919760 on Build GRCh38
rs778652125 Chr.11: 124919828-124919828 on Build GRCh38
rs780021223 Chr.11: 124919844-124919844 on Build GRCh38
rs746947486 Chr.11: 124919846-124919846 on Build GRCh38

View Full Product Details