Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 139356940-139357187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPB2
Entrez Gene ID
9276
Gene Name
coatomer protein complex subunit beta 2
Gene Aliases
beta'-COP
UniGene
Hs.681616
Gene Chromosome Location
Chr.3: 139355595-139389680 on Build GRCh38
Transcripts
XM_017007512.1 XM_017007513.1 NM_020191.2 XM_005247640.2 XM_006713703.3 XM_011512995.2 XM_011512996.2

Gene Symbol
MRPS22
Entrez Gene ID
56945
Gene Name
mitochondrial ribosomal protein S22
Gene Aliases
C3orf5, COXPD5, GIBT, GK002, MRP-S22, RPMS22
UniGene
Hs.745001
Gene Chromosome Location
Chr.3: 139343994-139357129 on Build GRCh38
Transcripts
XM_017007512.1 XM_017007513.1 NM_020191.2 XM_005247640.2 XM_006713703.3 XM_011512995.2 XM_011512996.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6247121 c.1018G>T p.A340S Chr.3: 139356949-139356949 on Build GRCh38
6715481 c.1035A>G p.T345T Chr.3: 139356966-139356966 on Build GRCh38
1039095 c.1063C>T p.R355C Chr.3: 139356994-139356994 on Build GRCh38
3124271 c.1021T>A p.Y341N Chr.3: 139356952-139356952 on Build GRCh38
107602 c.1014G>A p.K338K Chr.3: 139356945-139356945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986827742 Chr.3: 139357085-139357086 on Build GRCh38
rs556643495 Chr.3: 139357104-139357105 on Build GRCh38
rs886216431 Chr.3: 139356987-139356987 on Build GRCh38
rs764349006 Chr.3: 139357052-139357054 on Build GRCh38
rs755925113 Chr.3: 139356995-139356995 on Build GRCh38
rs15155 Chr.3: 139357021-139357021 on Build GRCh38
rs779498227 Chr.3: 139356969-139356969 on Build GRCh38
rs375619422 Chr.3: 139357032-139357032 on Build GRCh38
rs776389540 Chr.3: 139357065-139357066 on Build GRCh38
rs7833 Chr.3: 139357096-139357096 on Build GRCh38
rs191025290 Chr.3: 139356994-139356994 on Build GRCh38
rs1039665874 Chr.3: 139357092-139357092 on Build GRCh38
rs775727923 Chr.3: 139357068-139357068 on Build GRCh38
rs1056777532 Chr.3: 139356971-139356971 on Build GRCh38
rs886058024 Chr.3: 139356976-139356976 on Build GRCh38
rs765521114 Chr.3: 139357142-139357143 on Build GRCh38
rs956146100 Chr.3: 139357106-139357106 on Build GRCh38
rs980406032 Chr.3: 139357143-139357143 on Build GRCh38
rs887208171 Chr.3: 139357110-139357110 on Build GRCh38
rs74365525 Chr.3: 139357107-139357107 on Build GRCh38
rs1052650879 Chr.3: 139357011-139357011 on Build GRCh38
rs140638719 Chr.3: 139357069-139357069 on Build GRCh38
rs780864664 Chr.3: 139356992-139356992 on Build GRCh38
rs183391521 Chr.3: 139357116-139357116 on Build GRCh38
rs370173146 Chr.3: 139357034-139357034 on Build GRCh38
rs927227768 Chr.3: 139357157-139357158 on Build GRCh38
rs761161344 Chr.3: 139356976-139356977 on Build GRCh38
rs777482542 Chr.3: 139357024-139357024 on Build GRCh38
rs751069177 Chr.3: 139356978-139356978 on Build GRCh38
rs774350893 Chr.3: 139357053-139357053 on Build GRCh38
rs998278029 Chr.3: 139357105-139357105 on Build GRCh38
rs566795555 Chr.3: 139356991-139356991 on Build GRCh38
rs536270749 Chr.3: 139357162-139357162 on Build GRCh38
rs1007453766 Chr.3: 139357071-139357071 on Build GRCh38
rs80049554 Chr.3: 139357161-139357161 on Build GRCh38
rs946507268 Chr.3: 139357057-139357057 on Build GRCh38
rs770894198 Chr.3: 139357040-139357040 on Build GRCh38

View Full Product Details