Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70809792-70810039 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYDIN
Entrez Gene ID
54768
Gene Name
HYDIN, axonemal central pair apparatus protein
Gene Aliases
CILD5, HYDIN1, HYDIN2, PPP1R31
UniGene
Hs.351914
Gene Chromosome Location
Chr.16: 70802084-71230722 on Build GRCh38
Transcripts
NM_001270974.2 XM_006721206.2 XM_011523146.2 XM_011523147.1 XM_011523148.1 XM_011523151.1 XM_011523152.1 XM_011523155.2 XM_017023346.1 XM_017023347.1 XM_017023348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4691618 c.14638G>T p.E4880* Chr.16: 70809881-70809881 on Build GRCh38
4527503 c.14548G>A p.G4850R Chr.16: 70809971-70809971 on Build GRCh38
3795078 c.14605G>A p.E4869K Chr.16: 70809914-70809914 on Build GRCh38
327447 c.14638G>A p.E4880K Chr.16: 70809881-70809881 on Build GRCh38
3273461 c.14549G>T p.G4850V Chr.16: 70809970-70809970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986403779 Chr.16: 70809907-70809907 on Build GRCh38
rs748673208 Chr.16: 70809933-70809933 on Build GRCh38
rs368352014 Chr.16: 70809882-70809882 on Build GRCh38
rs374534223 Chr.16: 70809871-70809871 on Build GRCh38
rs373142549 Chr.16: 70809834-70809834 on Build GRCh38
rs1044250307 Chr.16: 70810007-70810007 on Build GRCh38
rs750579852 Chr.16: 70809891-70809891 on Build GRCh38
rs779331653 Chr.16: 70809929-70809929 on Build GRCh38
rs371130325 Chr.16: 70809844-70809844 on Build GRCh38
rs749735559 Chr.16: 70809830-70809830 on Build GRCh38
rs199704165 Chr.16: 70810006-70810006 on Build GRCh38
rs760419336 Chr.16: 70809857-70809857 on Build GRCh38
rs763025580 Chr.16: 70809998-70809998 on Build GRCh38
rs780507110 Chr.16: 70809814-70809814 on Build GRCh38
rs773845057 Chr.16: 70809836-70809836 on Build GRCh38
rs560482705 Chr.16: 70809958-70809958 on Build GRCh38
rs532437784 Chr.16: 70810001-70810001 on Build GRCh38
rs747587965 Chr.16: 70809840-70809840 on Build GRCh38
rs760801359 Chr.16: 70810003-70810003 on Build GRCh38
rs755338143 Chr.16: 70809904-70809904 on Build GRCh38
rs775528606 Chr.16: 70809988-70809988 on Build GRCh38
rs756394428 Chr.16: 70809813-70809813 on Build GRCh38
rs756408083 Chr.16: 70809894-70809894 on Build GRCh38
rs759348210 Chr.16: 70809978-70809978 on Build GRCh38
rs767737994 Chr.16: 70809890-70809890 on Build GRCh38
rs367919415 Chr.16: 70809960-70809960 on Build GRCh38
rs776551217 Chr.16: 70809866-70809866 on Build GRCh38
rs370629291 Chr.16: 70809883-70809883 on Build GRCh38
rs754293008 Chr.16: 70809899-70809899 on Build GRCh38
rs780267369 Chr.16: 70809895-70809895 on Build GRCh38
rs374038588 Chr.16: 70809959-70809959 on Build GRCh38
rs769734834 Chr.16: 70809987-70809987 on Build GRCh38
rs759390020 Chr.16: 70809868-70809868 on Build GRCh38
rs914971759 Chr.16: 70809980-70809981 on Build GRCh38
rs191626208 Chr.16: 70810002-70810002 on Build GRCh38
rs1018851502 Chr.16: 70809917-70809917 on Build GRCh38
rs776155283 Chr.16: 70809975-70809975 on Build GRCh38
rs766069657 Chr.16: 70809864-70809864 on Build GRCh38
rs771526729 Chr.16: 70809943-70809943 on Build GRCh38
rs772687296 Chr.16: 70809846-70809846 on Build GRCh38

View Full Product Details