Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151982941-151983214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNTL5
Entrez Gene ID
168391
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 5
Gene Aliases
GALNACT19, GALNT15, GalNAc-T5L
UniGene
Hs.647077
Gene Chromosome Location
Chr.7: 151956379-152019934 on Build GRCh38
Transcripts
NM_145292.3 NR_033169.1 XM_006715862.3 XM_011515849.2 XM_011515852.2 XM_011515853.2 XM_017011793.1 XM_017011794.1 XM_017011795.1 XM_017011796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1733379 c.394C>T p.R132C Chr.7: 151983011-151983011 on Build GRCh38
5997539 c.437A>G p.E146G Chr.7: 151983054-151983054 on Build GRCh38
4550537 c.502G>A p.E168K Chr.7: 151983119-151983119 on Build GRCh38
3262312 c.436G>T p.E146* Chr.7: 151983053-151983053 on Build GRCh38
7369183 c.370T>C p.C124R Chr.7: 151982987-151982987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6960270 Chr.7: 151982987-151982987 on Build GRCh38
rs769699726 Chr.7: 151983073-151983073 on Build GRCh38
rs141977214 Chr.7: 151982977-151982977 on Build GRCh38
rs182110483 Chr.7: 151983018-151983018 on Build GRCh38
rs756111158 Chr.7: 151983155-151983155 on Build GRCh38
rs776077983 Chr.7: 151983185-151983185 on Build GRCh38
rs370337705 Chr.7: 151982978-151982978 on Build GRCh38
rs369140074 Chr.7: 151982981-151982981 on Build GRCh38
rs981293137 Chr.7: 151983177-151983177 on Build GRCh38
rs35682935 Chr.7: 151983090-151983090 on Build GRCh38
rs765645704 Chr.7: 151983028-151983028 on Build GRCh38
rs868523131 Chr.7: 151983054-151983055 on Build GRCh38
rs377411415 Chr.7: 151983014-151983014 on Build GRCh38
rs989075318 Chr.7: 151983004-151983004 on Build GRCh38
rs560274426 Chr.7: 151983186-151983187 on Build GRCh38
rs777209665 Chr.7: 151982986-151982986 on Build GRCh38
rs756743311 Chr.7: 151983145-151983145 on Build GRCh38
rs147547437 Chr.7: 151983184-151983186 on Build GRCh38
rs765805106 Chr.7: 151983115-151983115 on Build GRCh38
rs758781978 Chr.7: 151983118-151983118 on Build GRCh38
rs766808347 Chr.7: 151983019-151983019 on Build GRCh38
rs200637744 Chr.7: 151983186-151983186 on Build GRCh38
rs568960098 Chr.7: 151982993-151982993 on Build GRCh38
rs768430205 Chr.7: 151983159-151983159 on Build GRCh38
rs897215657 Chr.7: 151983074-151983074 on Build GRCh38
rs781025768 Chr.7: 151983063-151983063 on Build GRCh38
rs753085976 Chr.7: 151983031-151983031 on Build GRCh38
rs201590087 Chr.7: 151983158-151983158 on Build GRCh38
rs150508704 Chr.7: 151983094-151983094 on Build GRCh38
rs776887374 Chr.7: 151983179-151983179 on Build GRCh38
rs200812489 Chr.7: 151983160-151983160 on Build GRCh38
rs113960815 Chr.7: 151983153-151983153 on Build GRCh38
rs745759083 Chr.7: 151983066-151983066 on Build GRCh38
rs758161025 Chr.7: 151983034-151983034 on Build GRCh38
rs575727995 Chr.7: 151983163-151983163 on Build GRCh38
rs35629905 Chr.7: 151983045-151983045 on Build GRCh38
rs756723086 Chr.7: 151983057-151983057 on Build GRCh38
rs957132442 Chr.7: 151983128-151983128 on Build GRCh38
rs137995289 Chr.7: 151983078-151983078 on Build GRCh38
rs777297966 Chr.7: 151983099-151983099 on Build GRCh38
rs759353909 Chr.7: 151982996-151982996 on Build GRCh38
rs769508887 Chr.7: 151983000-151983000 on Build GRCh38
rs948649134 Chr.7: 151983164-151983164 on Build GRCh38
rs755188105 Chr.7: 151983183-151983184 on Build GRCh38
rs947600384 Chr.7: 151983009-151983009 on Build GRCh38
rs770477529 Chr.7: 151983184-151983184 on Build GRCh38
rs537424966 Chr.7: 151983006-151983006 on Build GRCh38
rs780234715 Chr.7: 151983055-151983056 on Build GRCh38
rs199622614 Chr.7: 151983011-151983011 on Build GRCh38
rs760166425 Chr.7: 151983100-151983100 on Build GRCh38
rs771098985 Chr.7: 151982988-151982988 on Build GRCh38
rs777473316 Chr.7: 151983043-151983043 on Build GRCh38
rs771034600 Chr.7: 151983171-151983171 on Build GRCh38
rs766440773 Chr.7: 151983092-151983092 on Build GRCh38
rs763661672 Chr.7: 151983122-151983122 on Build GRCh38
rs372699787 Chr.7: 151983012-151983012 on Build GRCh38
rs752005387 Chr.7: 151983091-151983091 on Build GRCh38
rs370484975 Chr.7: 151983075-151983075 on Build GRCh38
rs751187784 Chr.7: 151983133-151983133 on Build GRCh38
rs35036517 Chr.7: 151983134-151983134 on Build GRCh38
rs764985593 Chr.7: 151983025-151983025 on Build GRCh38
rs202056038 Chr.7: 151983117-151983117 on Build GRCh38
rs772240728 Chr.7: 151983086-151983086 on Build GRCh38
rs755347291 Chr.7: 151982975-151982975 on Build GRCh38
rs544395807 Chr.7: 151983182-151983182 on Build GRCh38

View Full Product Details