Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50319677-50319924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYAL2
Entrez Gene ID
8692
Gene Name
hyaluronoglucosaminidase 2
Gene Aliases
LUCA2
UniGene
Hs.76873
Gene Chromosome Location
Chr.3: 50317790-50322850 on Build GRCh38
Transcripts
NM_003773.4 NM_033158.4 XM_005265524.2 XM_005265525.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1731080 c.593G>T p.R198L Chr.3: 50319897-50319897 on Build GRCh38
3595565 c.748C>T p.P250S Chr.3: 50319742-50319742 on Build GRCh38
5355365 c.765C>T p.D255D Chr.3: 50319725-50319725 on Build GRCh38
1566717 c.574C>T p.R192C Chr.3: 50319916-50319916 on Build GRCh38
6683283 c.811C>T p.R271C Chr.3: 50319679-50319679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050074 Chr.3: 50319705-50319705 on Build GRCh38
rs781990569 Chr.3: 50319856-50319856 on Build GRCh38
rs200942332 Chr.3: 50319725-50319725 on Build GRCh38
rs782751000 Chr.3: 50319863-50319863 on Build GRCh38
rs782819814 Chr.3: 50319769-50319769 on Build GRCh38
rs781966445 Chr.3: 50319821-50319821 on Build GRCh38
rs782762077 Chr.3: 50319793-50319793 on Build GRCh38
rs200201922 Chr.3: 50319753-50319753 on Build GRCh38
rs904074059 Chr.3: 50319738-50319738 on Build GRCh38
rs782055915 Chr.3: 50319756-50319756 on Build GRCh38
rs372371788 Chr.3: 50319789-50319789 on Build GRCh38
rs782410291 Chr.3: 50319902-50319902 on Build GRCh38
rs782511542 Chr.3: 50319706-50319706 on Build GRCh38
rs984721456 Chr.3: 50319871-50319871 on Build GRCh38
rs587616389 Chr.3: 50319891-50319891 on Build GRCh38
rs781914409 Chr.3: 50319734-50319734 on Build GRCh38
rs1031628155 Chr.3: 50319758-50319758 on Build GRCh38
rs78532114 Chr.3: 50319899-50319899 on Build GRCh38
rs782776218 Chr.3: 50319846-50319846 on Build GRCh38
rs782672655 Chr.3: 50319812-50319812 on Build GRCh38
rs782622084 Chr.3: 50319889-50319889 on Build GRCh38
rs1050073 Chr.3: 50319709-50319709 on Build GRCh38
rs782680699 Chr.3: 50319702-50319702 on Build GRCh38
rs375930599 Chr.3: 50319886-50319886 on Build GRCh38
rs781891669 Chr.3: 50319786-50319786 on Build GRCh38
rs782201493 Chr.3: 50319828-50319828 on Build GRCh38
rs35098416 Chr.3: 50319832-50319832 on Build GRCh38
rs782398975 Chr.3: 50319740-50319740 on Build GRCh38
rs782534238 Chr.3: 50319792-50319792 on Build GRCh38
rs782462647 Chr.3: 50319801-50319801 on Build GRCh38
rs149978009 Chr.3: 50319752-50319752 on Build GRCh38
rs782463118 Chr.3: 50319898-50319898 on Build GRCh38
rs146558025 Chr.3: 50319842-50319842 on Build GRCh38
rs368865414 Chr.3: 50319727-50319727 on Build GRCh38
rs782222986 Chr.3: 50319859-50319859 on Build GRCh38
rs782378463 Chr.3: 50319816-50319816 on Build GRCh38
rs781999115 Chr.3: 50319741-50319741 on Build GRCh38
rs371625877 Chr.3: 50319869-50319869 on Build GRCh38
rs11552691 Chr.3: 50319824-50319824 on Build GRCh38
rs782312446 Chr.3: 50319829-50319829 on Build GRCh38
rs782301565 Chr.3: 50319900-50319900 on Build GRCh38
rs781930973 Chr.3: 50319897-50319897 on Build GRCh38
rs781849879 Chr.3: 50319800-50319800 on Build GRCh38
rs368064171 Chr.3: 50319865-50319865 on Build GRCh38
rs782527322 Chr.3: 50319888-50319888 on Build GRCh38
rs184202689 Chr.3: 50319737-50319737 on Build GRCh38
rs996763404 Chr.3: 50319822-50319822 on Build GRCh38
rs782302544 Chr.3: 50319892-50319892 on Build GRCh38
rs892618784 Chr.3: 50319783-50319783 on Build GRCh38
rs374137877 Chr.3: 50319813-50319813 on Build GRCh38
rs782577291 Chr.3: 50319802-50319802 on Build GRCh38
rs782178070 Chr.3: 50319804-50319804 on Build GRCh38
rs199656869 Chr.3: 50319814-50319814 on Build GRCh38
rs782547130 Chr.3: 50319809-50319809 on Build GRCh38
rs781956056 Chr.3: 50319830-50319830 on Build GRCh38

View Full Product Details