Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151507475-151507702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT2
Entrez Gene ID
2196
Gene Name
FAT atypical cadherin 2
Gene Aliases
CDHF8, CDHR9, HFAT2, MEGF1
UniGene
Hs.591255
Gene Chromosome Location
Chr.5: 151504092-151594822 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Gene Symbol
SLC36A1
Entrez Gene ID
206358
Gene Name
solute carrier family 36 member 1
Gene Aliases
Dct1, LYAAT1, PAT1, TRAMD3
UniGene
Hs.269004
Gene Chromosome Location
Chr.5: 151411344-151556318 on Build GRCh38
Transcripts
NM_001447.2 XM_006714761.3 XM_011537600.2 XM_011537603.2 XM_017009224.1 XM_017009225.1 XM_011537595.2 XM_017009217.1 XM_017009219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660868 c.12191T>C p.V4064A Chr.5: 151507480-151507480 on Build GRCh38
6583005 c.12070G>T p.E4024* Chr.5: 151507601-151507601 on Build GRCh38
7266450 c.12074C>T p.A4025V Chr.5: 151507597-151507597 on Build GRCh38
3852886 c.12165C>T p.A4055A Chr.5: 151507506-151507506 on Build GRCh38
1718827 c.12164C>T p.A4055V Chr.5: 151507507-151507507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369964992 Chr.5: 151507594-151507594 on Build GRCh38
rs983973270 Chr.5: 151507667-151507667 on Build GRCh38
rs376838866 Chr.5: 151507634-151507634 on Build GRCh38
rs760967696 Chr.5: 151507506-151507506 on Build GRCh38
rs759144402 Chr.5: 151507580-151507582 on Build GRCh38
rs894736376 Chr.5: 151507569-151507569 on Build GRCh38
rs528067490 Chr.5: 151507547-151507547 on Build GRCh38
rs900550864 Chr.5: 151507580-151507580 on Build GRCh38
rs752136554 Chr.5: 151507631-151507632 on Build GRCh38
rs143949083 Chr.5: 151507505-151507505 on Build GRCh38
rs80113300 Chr.5: 151507626-151507626 on Build GRCh38
rs748636534 Chr.5: 151507616-151507616 on Build GRCh38
rs147299560 Chr.5: 151507512-151507512 on Build GRCh38
rs373396929 Chr.5: 151507598-151507598 on Build GRCh38
rs201790770 Chr.5: 151507615-151507615 on Build GRCh38
rs66498388 Chr.5: 151507624-151507625 on Build GRCh38
rs538316910 Chr.5: 151507597-151507597 on Build GRCh38
rs761057720 Chr.5: 151507596-151507596 on Build GRCh38
rs372619398 Chr.5: 151507645-151507645 on Build GRCh38
rs1021434484 Chr.5: 151507515-151507515 on Build GRCh38
rs201517523 Chr.5: 151507508-151507508 on Build GRCh38
rs75548276 Chr.5: 151507623-151507624 on Build GRCh38
rs760094674 Chr.5: 151507583-151507583 on Build GRCh38
rs770302828 Chr.5: 151507571-151507571 on Build GRCh38
rs764457790 Chr.5: 151507630-151507630 on Build GRCh38
rs772761270 Chr.5: 151507591-151507591 on Build GRCh38
rs767959528 Chr.5: 151507501-151507501 on Build GRCh38
rs775632768 Chr.5: 151507544-151507544 on Build GRCh38
rs748991728 Chr.5: 151507560-151507560 on Build GRCh38
rs748857819 Chr.5: 151507638-151507638 on Build GRCh38
rs201804836 Chr.5: 151507548-151507548 on Build GRCh38
rs56258915 Chr.5: 151507625-151507626 on Build GRCh38
rs150543833 Chr.5: 151507601-151507601 on Build GRCh38
rs867210819 Chr.5: 151507502-151507502 on Build GRCh38
rs148911050 Chr.5: 151507665-151507665 on Build GRCh38
rs41290577 Chr.5: 151507639-151507639 on Build GRCh38
rs568706541 Chr.5: 151507636-151507636 on Build GRCh38
rs138553780 Chr.5: 151507500-151507500 on Build GRCh38
rs750553614 Chr.5: 151507612-151507612 on Build GRCh38
rs752643829 Chr.5: 151507631-151507631 on Build GRCh38
rs138750878 Chr.5: 151507559-151507559 on Build GRCh38
rs140966881 Chr.5: 151507524-151507524 on Build GRCh38
rs745440377 Chr.5: 151507655-151507655 on Build GRCh38

View Full Product Details