Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51746662-51746935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPR1
Entrez Gene ID
2357
Gene Name
formyl peptide receptor 1
Gene Aliases
FMLP, FPR
UniGene
Hs.753
Gene Chromosome Location
Chr.19: 51745770-51751897 on Build GRCh38
Transcripts
NM_001193306.1 NM_002029.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6151729 c.274C>G p.P92A Chr.19: 51746721-51746721 on Build GRCh38
1494334 c.161G>A p.R54Q Chr.19: 51746834-51746834 on Build GRCh38
5554345 c.240C>A p.F80L Chr.19: 51746755-51746755 on Build GRCh38
6525287 c.273G>T p.W91C Chr.19: 51746722-51746722 on Build GRCh38
5667551 c.268C>A p.H90N Chr.19: 51746727-51746727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759165021 Chr.19: 51746856-51746856 on Build GRCh38
rs373383845 Chr.19: 51746722-51746722 on Build GRCh38
rs1039474132 Chr.19: 51746909-51746909 on Build GRCh38
rs767155719 Chr.19: 51746857-51746857 on Build GRCh38
rs752038261 Chr.19: 51746717-51746717 on Build GRCh38
rs780182759 Chr.19: 51746894-51746894 on Build GRCh38
rs754232242 Chr.19: 51746760-51746760 on Build GRCh38
rs1013579949 Chr.19: 51746880-51746880 on Build GRCh38
rs753486789 Chr.19: 51746903-51746904 on Build GRCh38
rs374349320 Chr.19: 51746716-51746716 on Build GRCh38
rs199764870 Chr.19: 51746695-51746695 on Build GRCh38
rs375049372 Chr.19: 51746733-51746733 on Build GRCh38
rs754359145 Chr.19: 51746862-51746862 on Build GRCh38
rs747128724 Chr.19: 51746809-51746809 on Build GRCh38
rs765852801 Chr.19: 51746724-51746724 on Build GRCh38
rs764850416 Chr.19: 51746710-51746710 on Build GRCh38
rs1015598098 Chr.19: 51746709-51746709 on Build GRCh38
rs773879518 Chr.19: 51746852-51746852 on Build GRCh38
rs148846448 Chr.19: 51746735-51746735 on Build GRCh38
rs143480239 Chr.19: 51746791-51746791 on Build GRCh38
rs145808420 Chr.19: 51746834-51746834 on Build GRCh38
rs764738404 Chr.19: 51746750-51746750 on Build GRCh38
rs756639389 Chr.19: 51746730-51746730 on Build GRCh38
rs765639685 Chr.19: 51746876-51746876 on Build GRCh38
rs748109117 Chr.19: 51746829-51746829 on Build GRCh38
rs78488639 Chr.19: 51746706-51746706 on Build GRCh38
rs750324912 Chr.19: 51746721-51746721 on Build GRCh38
rs138475342 Chr.19: 51746835-51746835 on Build GRCh38
rs1044585176 Chr.19: 51746902-51746902 on Build GRCh38
rs770552297 Chr.19: 51746850-51746850 on Build GRCh38
rs767877982 Chr.19: 51746859-51746865 on Build GRCh38
rs751640032 Chr.19: 51746904-51746904 on Build GRCh38
rs1023206431 Chr.19: 51746779-51746779 on Build GRCh38
rs376354678 Chr.19: 51746725-51746725 on Build GRCh38
rs199534146 Chr.19: 51746790-51746790 on Build GRCh38
rs28930680 Chr.19: 51746689-51746689 on Build GRCh38
rs762650605 Chr.19: 51746844-51746844 on Build GRCh38
rs371282377 Chr.19: 51746797-51746797 on Build GRCh38
rs768554070 Chr.19: 51746810-51746810 on Build GRCh38
rs952836159 Chr.19: 51746697-51746697 on Build GRCh38
rs750811701 Chr.19: 51746877-51746877 on Build GRCh38
rs149382837 Chr.19: 51746878-51746878 on Build GRCh38
rs768642242 Chr.19: 51746698-51746698 on Build GRCh38
rs148014099 Chr.19: 51746806-51746806 on Build GRCh38
rs762415254 Chr.19: 51746863-51746863 on Build GRCh38
rs910217049 Chr.19: 51746803-51746803 on Build GRCh38
rs61736491 Chr.19: 51746818-51746818 on Build GRCh38
rs757595295 Chr.19: 51746722-51746722 on Build GRCh38
rs866198922 Chr.19: 51746714-51746714 on Build GRCh38
rs2070745 Chr.19: 51746694-51746694 on Build GRCh38

View Full Product Details