Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30905174-30905421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAN1
Entrez Gene ID
22909
Gene Name
FANCD2/FANCI-associated nuclease 1
Gene Aliases
KIAA1018, KMIN, MTMR15, hFAN1
UniGene
Hs.584863
Gene Chromosome Location
Chr.15: 30903695-30943108 on Build GRCh38
Transcripts
NM_001146094.1 NM_001146095.1 NM_001146096.1 NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_011521372.2 XM_017022012.1 XM_017022013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
700209 c.530C>T p.A177V Chr.15: 30905193-30905193 on Build GRCh38
6351183 c.557C>A p.S186Y Chr.15: 30905220-30905220 on Build GRCh38
1177709 c.686A>G p.H229R Chr.15: 30905349-30905349 on Build GRCh38
6351182 c.557C>A p.S186Y Chr.15: 30905220-30905220 on Build GRCh38
4683332 c.642C>T p.N214N Chr.15: 30905305-30905305 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189713653 Chr.15: 30905319-30905319 on Build GRCh38
rs1023915200 Chr.15: 30905373-30905373 on Build GRCh38
rs750997311 Chr.15: 30905350-30905350 on Build GRCh38
rs982267047 Chr.15: 30905222-30905222 on Build GRCh38
rs774210518 Chr.15: 30905398-30905398 on Build GRCh38
rs150748572 Chr.15: 30905381-30905381 on Build GRCh38
rs148140498 Chr.15: 30905348-30905348 on Build GRCh38
rs747840189 Chr.15: 30905265-30905265 on Build GRCh38
rs1040089394 Chr.15: 30905345-30905345 on Build GRCh38
rs770745828 Chr.15: 30905395-30905395 on Build GRCh38
rs767736939 Chr.15: 30905387-30905387 on Build GRCh38
rs143816528 Chr.15: 30905318-30905318 on Build GRCh38
rs768300893 Chr.15: 30905248-30905248 on Build GRCh38
rs201722958 Chr.15: 30905236-30905236 on Build GRCh38
rs771480956 Chr.15: 30905315-30905315 on Build GRCh38
rs770905269 Chr.15: 30905307-30905307 on Build GRCh38
rs754469574 Chr.15: 30905353-30905353 on Build GRCh38
rs774654411 Chr.15: 30905319-30905327 on Build GRCh38
rs369869010 Chr.15: 30905335-30905335 on Build GRCh38
rs920551872 Chr.15: 30905352-30905352 on Build GRCh38
rs376528017 Chr.15: 30905329-30905329 on Build GRCh38
rs376369585 Chr.15: 30905253-30905253 on Build GRCh38
rs377089680 Chr.15: 30905228-30905228 on Build GRCh38
rs749848190 Chr.15: 30905238-30905241 on Build GRCh38
rs200798913 Chr.15: 30905394-30905394 on Build GRCh38
rs773116536 Chr.15: 30905267-30905267 on Build GRCh38
rs369093658 Chr.15: 30905261-30905261 on Build GRCh38
rs759574299 Chr.15: 30905325-30905325 on Build GRCh38
rs547265978 Chr.15: 30905374-30905374 on Build GRCh38
rs779675652 Chr.15: 30905217-30905217 on Build GRCh38
rs762769712 Chr.15: 30905294-30905294 on Build GRCh38
rs780766743 Chr.15: 30905359-30905359 on Build GRCh38
rs774296372 Chr.15: 30905315-30905315 on Build GRCh38
rs897593551 Chr.15: 30905347-30905347 on Build GRCh38
rs148610242 Chr.15: 30905220-30905220 on Build GRCh38
rs757813353 Chr.15: 30905349-30905349 on Build GRCh38
rs778218138 Chr.15: 30905204-30905204 on Build GRCh38
rs754266102 Chr.15: 30905342-30905342 on Build GRCh38
rs757901655 Chr.15: 30905215-30905215 on Build GRCh38
rs751203175 Chr.15: 30905369-30905369 on Build GRCh38
rs374878731 Chr.15: 30905314-30905314 on Build GRCh38
rs754291086 Chr.15: 30905208-30905208 on Build GRCh38
rs761020052 Chr.15: 30905336-30905336 on Build GRCh38
rs142084532 Chr.15: 30905266-30905266 on Build GRCh38
rs373390525 Chr.15: 30905271-30905271 on Build GRCh38
rs745822183 Chr.15: 30905399-30905399 on Build GRCh38
rs770015167 Chr.15: 30905288-30905288 on Build GRCh38
rs17846417 Chr.15: 30905364-30905364 on Build GRCh38
rs772700881 Chr.15: 30905221-30905222 on Build GRCh38
rs764365598 Chr.15: 30905339-30905339 on Build GRCh38

View Full Product Details