Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94067535-94067790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEPHL1
Entrez Gene ID
341208
Gene Name
hephaestin like 1
Gene Aliases
-
UniGene
Hs.669645
Gene Chromosome Location
Chr.11: 94021212-94114208 on Build GRCh38
Transcripts
NM_001098672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
690927 c.871T>A p.S291T Chr.11: 94067558-94067558 on Build GRCh38
6552562 c.928G>T p.G310C Chr.11: 94067615-94067615 on Build GRCh38
4690010 c.1019C>T p.P340L Chr.11: 94067706-94067706 on Build GRCh38
4514528 c.978C>T p.F326F Chr.11: 94067665-94067665 on Build GRCh38
4512830 c.918C>T p.I306I Chr.11: 94067605-94067605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778594960 Chr.11: 94067721-94067721 on Build GRCh38
rs369122858 Chr.11: 94067668-94067668 on Build GRCh38
rs754809301 Chr.11: 94067651-94067653 on Build GRCh38
rs920009521 Chr.11: 94067579-94067579 on Build GRCh38
rs1053915084 Chr.11: 94067678-94067678 on Build GRCh38
rs893566464 Chr.11: 94067682-94067682 on Build GRCh38
rs375642305 Chr.11: 94067699-94067699 on Build GRCh38
rs537000260 Chr.11: 94067603-94067603 on Build GRCh38
rs774463623 Chr.11: 94067750-94067750 on Build GRCh38
rs921464921 Chr.11: 94067658-94067658 on Build GRCh38
rs755770180 Chr.11: 94067566-94067566 on Build GRCh38
rs778772903 Chr.11: 94067644-94067644 on Build GRCh38
rs746280308 Chr.11: 94067737-94067737 on Build GRCh38
rs763194956 Chr.11: 94067743-94067743 on Build GRCh38
rs759918213 Chr.11: 94067640-94067640 on Build GRCh38
rs766006698 Chr.11: 94067600-94067603 on Build GRCh38
rs779629055 Chr.11: 94067698-94067698 on Build GRCh38
rs61220720 Chr.11: 94067584-94067584 on Build GRCh38
rs765449710 Chr.11: 94067642-94067642 on Build GRCh38
rs771425413 Chr.11: 94067628-94067628 on Build GRCh38
rs983197066 Chr.11: 94067611-94067611 on Build GRCh38
rs541253114 Chr.11: 94067666-94067666 on Build GRCh38
rs776878491 Chr.11: 94067636-94067636 on Build GRCh38
rs750359125 Chr.11: 94067758-94067758 on Build GRCh38
rs775768088 Chr.11: 94067742-94067742 on Build GRCh38
rs373413432 Chr.11: 94067684-94067684 on Build GRCh38
rs371190382 Chr.11: 94067728-94067728 on Build GRCh38
rs932802377 Chr.11: 94067677-94067677 on Build GRCh38
rs745627455 Chr.11: 94067562-94067562 on Build GRCh38
rs373013506 Chr.11: 94067653-94067653 on Build GRCh38
rs375991157 Chr.11: 94067657-94067657 on Build GRCh38
rs556037808 Chr.11: 94067613-94067613 on Build GRCh38
rs775678346 Chr.11: 94067643-94067643 on Build GRCh38
rs781721933 Chr.11: 94067735-94067735 on Build GRCh38
rs761921362 Chr.11: 94067757-94067757 on Build GRCh38
rs183813405 Chr.11: 94067654-94067654 on Build GRCh38
rs747521269 Chr.11: 94067622-94067622 on Build GRCh38
rs751399244 Chr.11: 94067608-94067608 on Build GRCh38
rs747609253 Chr.11: 94067724-94067724 on Build GRCh38
rs991569554 Chr.11: 94067564-94067564 on Build GRCh38
rs779725308 Chr.11: 94067596-94067596 on Build GRCh38
rs770211234 Chr.11: 94067738-94067738 on Build GRCh38
rs537253048 Chr.11: 94067705-94067705 on Build GRCh38
rs768817851 Chr.11: 94067745-94067745 on Build GRCh38

View Full Product Details