Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134359282-134359537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMOTL2
Entrez Gene ID
51421
Gene Name
angiomotin like 2
Gene Aliases
LCCP
UniGene
Hs.426312
Gene Chromosome Location
Chr.3: 134355345-134375417 on Build GRCh38
Transcripts
NM_001278683.1 NM_001278685.1 NM_016201.3 XM_006713654.2 XM_011512881.1 XM_017006580.1 XM_017006581.1 XM_017006582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4319293 c.1930A>G p.K644E Chr.3: 134359457-134359457 on Build GRCh38
7255905 c.2069G>T p.R690L Chr.3: 134359318-134359318 on Build GRCh38
1419223 c.2018C>T p.A673V Chr.3: 134359369-134359369 on Build GRCh38
5994685 c.1978G>T p.G660C Chr.3: 134359409-134359409 on Build GRCh38
3122638 c.2000C>T p.S667L Chr.3: 134359387-134359387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780310495 Chr.3: 134359315-134359315 on Build GRCh38
rs745539705 Chr.3: 134359363-134359363 on Build GRCh38
rs140943062 Chr.3: 134359332-134359332 on Build GRCh38
rs781488712 Chr.3: 134359400-134359400 on Build GRCh38
rs778598828 Chr.3: 134359409-134359409 on Build GRCh38
rs756779585 Chr.3: 134359406-134359406 on Build GRCh38
rs903909177 Chr.3: 134359381-134359381 on Build GRCh38
rs147292735 Chr.3: 134359305-134359305 on Build GRCh38
rs147485667 Chr.3: 134359441-134359441 on Build GRCh38
rs376138663 Chr.3: 134359371-134359371 on Build GRCh38
rs1043764172 Chr.3: 134359372-134359372 on Build GRCh38
rs781753734 Chr.3: 134359319-134359319 on Build GRCh38
rs142786862 Chr.3: 134359458-134359458 on Build GRCh38
rs756659217 Chr.3: 134359494-134359494 on Build GRCh38
rs766509546 Chr.3: 134359456-134359456 on Build GRCh38
rs775118618 Chr.3: 134359369-134359369 on Build GRCh38
rs997508151 Chr.3: 134359397-134359397 on Build GRCh38
rs368824669 Chr.3: 134359436-134359436 on Build GRCh38
rs557857902 Chr.3: 134359385-134359385 on Build GRCh38
rs751859743 Chr.3: 134359399-134359399 on Build GRCh38
rs751814161 Chr.3: 134359469-134359469 on Build GRCh38
rs750181224 Chr.3: 134359513-134359514 on Build GRCh38
rs767383888 Chr.3: 134359479-134359479 on Build GRCh38
rs577285464 Chr.3: 134359417-134359417 on Build GRCh38
rs372900931 Chr.3: 134359304-134359304 on Build GRCh38
rs962403760 Chr.3: 134359507-134359507 on Build GRCh38
rs770573770 Chr.3: 134359343-134359343 on Build GRCh38
rs760574540 Chr.3: 134359370-134359370 on Build GRCh38
rs753216504 Chr.3: 134359493-134359493 on Build GRCh38
rs747438671 Chr.3: 134359317-134359317 on Build GRCh38
rs746713717 Chr.3: 134359418-134359418 on Build GRCh38
rs749945908 Chr.3: 134359517-134359517 on Build GRCh38
rs778474459 Chr.3: 134359506-134359506 on Build GRCh38
rs189825181 Chr.3: 134359383-134359383 on Build GRCh38
rs771603095 Chr.3: 134359413-134359413 on Build GRCh38
rs778650330 Chr.3: 134359362-134359362 on Build GRCh38
rs750600683 Chr.3: 134359386-134359386 on Build GRCh38
rs543449878 Chr.3: 134359368-134359368 on Build GRCh38
rs759765618 Chr.3: 134359474-134359474 on Build GRCh38
rs980551415 Chr.3: 134359508-134359508 on Build GRCh38
rs1026597225 Chr.3: 134359452-134359452 on Build GRCh38
rs897608608 Chr.3: 134359329-134359329 on Build GRCh38
rs144685493 Chr.3: 134359387-134359387 on Build GRCh38
rs763153527 Chr.3: 134359444-134359444 on Build GRCh38
rs982390997 Chr.3: 134359471-134359471 on Build GRCh38
rs140029062 Chr.3: 134359442-134359442 on Build GRCh38
rs767947578 Chr.3: 134359488-134359488 on Build GRCh38
rs556572166 Chr.3: 134359318-134359318 on Build GRCh38
rs768509607 Chr.3: 134359434-134359434 on Build GRCh38
rs951033847 Chr.3: 134359467-134359467 on Build GRCh38
rs753273915 Chr.3: 134359405-134359405 on Build GRCh38
rs375617544 Chr.3: 134359382-134359382 on Build GRCh38
rs745411834 Chr.3: 134359411-134359411 on Build GRCh38

View Full Product Details