Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 103574207-103574476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMY2B
Entrez Gene ID
280
Gene Name
amylase, alpha 2B (pancreatic)
Gene Aliases
AMY2, AMY3, HXA
UniGene
Hs.484588
Gene Chromosome Location
Chr.1: 103554644-103579534 on Build GRCh38
Transcripts
NM_020978.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3801095 c.803T>G p.V268G Chr.1: 103574318-103574318 on Build GRCh38
6120226 c.756G>T p.L252L Chr.1: 103574271-103574271 on Build GRCh38
3801094 c.754C>T p.L252L Chr.1: 103574269-103574269 on Build GRCh38
4979556 c.852G>C p.W284C Chr.1: 103574367-103574367 on Build GRCh38
3356422 c.841A>T p.I281F Chr.1: 103574356-103574356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006956219 Chr.1: 103574442-103574442 on Build GRCh38
rs147458656 Chr.1: 103574382-103574382 on Build GRCh38
rs776893049 Chr.1: 103574259-103574259 on Build GRCh38
rs369145442 Chr.1: 103574307-103574307 on Build GRCh38
rs897113237 Chr.1: 103574362-103574362 on Build GRCh38
rs184979235 Chr.1: 103574371-103574371 on Build GRCh38
rs781566703 Chr.1: 103574383-103574383 on Build GRCh38
rs764330670 Chr.1: 103574281-103574281 on Build GRCh38
rs372593038 Chr.1: 103574375-103574375 on Build GRCh38
rs769935411 Chr.1: 103574400-103574400 on Build GRCh38
rs749359311 Chr.1: 103574423-103574423 on Build GRCh38
rs550691635 Chr.1: 103574352-103574352 on Build GRCh38
rs763325013 Chr.1: 103574346-103574346 on Build GRCh38
rs994172772 Chr.1: 103574393-103574393 on Build GRCh38
rs765833962 Chr.1: 103574273-103574273 on Build GRCh38
rs774829154 Chr.1: 103574434-103574434 on Build GRCh38
rs538994510 Chr.1: 103574339-103574339 on Build GRCh38
rs747919666 Chr.1: 103574321-103574321 on Build GRCh38
rs764566458 Chr.1: 103574347-103574347 on Build GRCh38
rs754910810 Chr.1: 103574373-103574373 on Build GRCh38
rs759532881 Chr.1: 103574266-103574266 on Build GRCh38
rs767804939 Chr.1: 103574438-103574438 on Build GRCh38
rs374395229 Chr.1: 103574314-103574314 on Build GRCh38
rs138821426 Chr.1: 103574343-103574343 on Build GRCh38
rs761902863 Chr.1: 103574398-103574399 on Build GRCh38
rs768277222 Chr.1: 103574242-103574242 on Build GRCh38
rs1048912032 Chr.1: 103574408-103574408 on Build GRCh38
rs775485072 Chr.1: 103574421-103574421 on Build GRCh38
rs762181012 Chr.1: 103574436-103574436 on Build GRCh38
rs763022449 Chr.1: 103574389-103574389 on Build GRCh38
rs200275225 Chr.1: 103574315-103574315 on Build GRCh38
rs201396892 Chr.1: 103574360-103574360 on Build GRCh38
rs746033188 Chr.1: 103574394-103574394 on Build GRCh38
rs771256614 Chr.1: 103574258-103574258 on Build GRCh38
rs750004107 Chr.1: 103574305-103574305 on Build GRCh38
rs555648662 Chr.1: 103574444-103574444 on Build GRCh38
rs150659982 Chr.1: 103574333-103574333 on Build GRCh38
rs12131520 Chr.1: 103574298-103574298 on Build GRCh38
rs752838004 Chr.1: 103574369-103574369 on Build GRCh38
rs769220789 Chr.1: 103574430-103574430 on Build GRCh38
rs370085323 Chr.1: 103574285-103574285 on Build GRCh38
rs199872830 Chr.1: 103574424-103574424 on Build GRCh38
rs751964563 Chr.1: 103574354-103574354 on Build GRCh38
rs771344389 Chr.1: 103574324-103574324 on Build GRCh38
rs778899651 Chr.1: 103574320-103574320 on Build GRCh38
rs748012161 Chr.1: 103574378-103574378 on Build GRCh38
rs763384035 Chr.1: 103574276-103574276 on Build GRCh38
rs762164525 Chr.1: 103574356-103574356 on Build GRCh38
rs768571213 Chr.1: 103574324-103574330 on Build GRCh38
rs776977215 Chr.1: 103574332-103574332 on Build GRCh38
rs368462934 Chr.1: 103574359-103574359 on Build GRCh38
rs776585530 Chr.1: 103574390-103574390 on Build GRCh38
rs753090470 Chr.1: 103574275-103574275 on Build GRCh38
rs193276867 Chr.1: 103574318-103574318 on Build GRCh38
rs965250538 Chr.1: 103574296-103574297 on Build GRCh38
rs773571986 Chr.1: 103574256-103574256 on Build GRCh38

View Full Product Details