Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96839496-96839735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD23
Entrez Gene ID
200539
Gene Name
ankyrin repeat domain 23
Gene Aliases
DARP, MARP3
UniGene
Hs.643430
Gene Chromosome Location
Chr.2: 96837914-96844021 on Build GRCh38
Transcripts
NM_144994.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4970265 c.897G>A p.A299A Chr.2: 96839570-96839570 on Build GRCh38
238785 c.911G>A p.R304Q Chr.2: 96839556-96839556 on Build GRCh38
4661803 c.910C>T p.R304W Chr.2: 96839557-96839557 on Build GRCh38
4985666 c.872G>C p.R291P Chr.2: 96839595-96839595 on Build GRCh38
6243610 c.829G>A p.V277M Chr.2: 96839638-96839638 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997096947 Chr.2: 96839657-96839657 on Build GRCh38
rs766760232 Chr.2: 96839618-96839618 on Build GRCh38
rs764548904 Chr.2: 96839578-96839578 on Build GRCh38
rs750593646 Chr.2: 96839603-96839603 on Build GRCh38
rs570323112 Chr.2: 96839565-96839565 on Build GRCh38
rs768125459 Chr.2: 96839673-96839673 on Build GRCh38
rs778794580 Chr.2: 96839634-96839634 on Build GRCh38
rs758362502 Chr.2: 96839643-96839643 on Build GRCh38
rs747550691 Chr.2: 96839608-96839609 on Build GRCh38
rs575883381 Chr.2: 96839712-96839712 on Build GRCh38
rs754691835 Chr.2: 96839631-96839631 on Build GRCh38
rs753064910 Chr.2: 96839516-96839516 on Build GRCh38
rs967214847 Chr.2: 96839571-96839571 on Build GRCh38
rs938164758 Chr.2: 96839709-96839709 on Build GRCh38
rs373941815 Chr.2: 96839656-96839656 on Build GRCh38
rs986068160 Chr.2: 96839617-96839617 on Build GRCh38
rs772606680 Chr.2: 96839689-96839689 on Build GRCh38
rs751627806 Chr.2: 96839527-96839527 on Build GRCh38
rs553761791 Chr.2: 96839665-96839665 on Build GRCh38
rs746580496 Chr.2: 96839649-96839649 on Build GRCh38
rs769105015 Chr.2: 96839718-96839719 on Build GRCh38
rs990115968 Chr.2: 96839537-96839537 on Build GRCh38
rs374745685 Chr.2: 96839693-96839693 on Build GRCh38
rs533331557 Chr.2: 96839563-96839563 on Build GRCh38
rs549596954 Chr.2: 96839614-96839614 on Build GRCh38
rs778759680 Chr.2: 96839561-96839561 on Build GRCh38
rs529514658 Chr.2: 96839604-96839604 on Build GRCh38
rs74826568 Chr.2: 96839560-96839560 on Build GRCh38
rs764557718 Chr.2: 96839521-96839521 on Build GRCh38
rs929045184 Chr.2: 96839549-96839549 on Build GRCh38
rs775808923 Chr.2: 96839567-96839567 on Build GRCh38
rs756407606 Chr.2: 96839547-96839547 on Build GRCh38
rs774867231 Chr.2: 96839596-96839597 on Build GRCh38
rs914788498 Chr.2: 96839545-96839545 on Build GRCh38
rs749149634 Chr.2: 96839551-96839551 on Build GRCh38
rs758257387 Chr.2: 96839706-96839706 on Build GRCh38
rs372499217 Chr.2: 96839715-96839715 on Build GRCh38
rs568204652 Chr.2: 96839648-96839648 on Build GRCh38
rs914567696 Chr.2: 96839696-96839696 on Build GRCh38
rs774778805 Chr.2: 96839668-96839668 on Build GRCh38
rs370360720 Chr.2: 96839664-96839664 on Build GRCh38
rs200849217 Chr.2: 96839562-96839562 on Build GRCh38
rs753658719 Chr.2: 96839625-96839625 on Build GRCh38
rs780034472 Chr.2: 96839713-96839713 on Build GRCh38
rs780693506 Chr.2: 96839719-96839719 on Build GRCh38
rs201677881 Chr.2: 96839663-96839663 on Build GRCh38
rs756707778 Chr.2: 96839717-96839717 on Build GRCh38
rs763448108 Chr.2: 96839520-96839520 on Build GRCh38
rs1019970684 Chr.2: 96839641-96839641 on Build GRCh38
rs766600523 Chr.2: 96839690-96839690 on Build GRCh38
rs547811122 Chr.2: 96839533-96839533 on Build GRCh38
rs370500398 Chr.2: 96839591-96839591 on Build GRCh38
rs747581640 Chr.2: 96839554-96839554 on Build GRCh38
rs750612151 Chr.2: 96839546-96839546 on Build GRCh38
rs752606484 Chr.2: 96839701-96839701 on Build GRCh38
rs944334849 Chr.2: 96839531-96839532 on Build GRCh38
rs771412496 Chr.2: 96839615-96839615 on Build GRCh38
rs747978980 Chr.2: 96839639-96839639 on Build GRCh38
rs539381940 Chr.2: 96839675-96839675 on Build GRCh38
rs201589615 Chr.2: 96839694-96839694 on Build GRCh38
rs570371939 Chr.2: 96839670-96839670 on Build GRCh38
rs550222955 Chr.2: 96839564-96839564 on Build GRCh38
rs757242343 Chr.2: 96839532-96839532 on Build GRCh38
rs774670824 Chr.2: 96839594-96839594 on Build GRCh38
rs922028521 Chr.2: 96839517-96839517 on Build GRCh38
rs1048087360 Chr.2: 96839597-96839597 on Build GRCh38
rs373064902 Chr.2: 96839519-96839519 on Build GRCh38

View Full Product Details