Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27057092-27057347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL5
Entrez Gene ID
60509
Gene Name
ATP/GTP binding protein like 5
Gene Aliases
CCP5, RP75
UniGene
Hs.138207
Gene Chromosome Location
Chr.2: 27051423-27070622 on Build GRCh38
Transcripts
NM_001035507.2 NM_021831.5 NR_104246.1 XM_005264477.2 XM_011533011.2 XM_011533012.2 XM_011533013.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4624777 c.1540A>G p.T514A Chr.2: 27057307-27057307 on Build GRCh38
1283309 c.1568G>A p.R523H Chr.2: 27057335-27057335 on Build GRCh38
3580491 c.1564G>A p.G522R Chr.2: 27057331-27057331 on Build GRCh38
1283310 c.1568G>A p.R523H Chr.2: 27057335-27057335 on Build GRCh38
4624776 c.1540A>G p.T514A Chr.2: 27057307-27057307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935122584 Chr.2: 27057136-27057136 on Build GRCh38
rs765084879 Chr.2: 27057309-27057309 on Build GRCh38
rs775621095 Chr.2: 27057124-27057125 on Build GRCh38
rs1009027661 Chr.2: 27057274-27057274 on Build GRCh38
rs750182899 Chr.2: 27057160-27057160 on Build GRCh38
rs6711384 Chr.2: 27057224-27057224 on Build GRCh38
rs370699569 Chr.2: 27057146-27057147 on Build GRCh38
rs1009000368 Chr.2: 27057187-27057187 on Build GRCh38
rs907627090 Chr.2: 27057169-27057169 on Build GRCh38
rs776566861 Chr.2: 27057291-27057291 on Build GRCh38
rs750275368 Chr.2: 27057312-27057312 on Build GRCh38
rs114892980 Chr.2: 27057253-27057253 on Build GRCh38
rs771329023 Chr.2: 27057262-27057262 on Build GRCh38
rs6721395 Chr.2: 27057259-27057259 on Build GRCh38
rs1053626491 Chr.2: 27057137-27057137 on Build GRCh38
rs568586485 Chr.2: 27057265-27057265 on Build GRCh38
rs899395892 Chr.2: 27057258-27057258 on Build GRCh38
rs754229032 Chr.2: 27057187-27057187 on Build GRCh38
rs931715968 Chr.2: 27057144-27057144 on Build GRCh38
rs890113182 Chr.2: 27057171-27057171 on Build GRCh38
rs143616306 Chr.2: 27057273-27057273 on Build GRCh38
rs545721507 Chr.2: 27057142-27057143 on Build GRCh38
rs374072681 Chr.2: 27057320-27057320 on Build GRCh38
rs55837250 Chr.2: 27057288-27057288 on Build GRCh38
rs755993908 Chr.2: 27057208-27057208 on Build GRCh38
rs192886059 Chr.2: 27057240-27057240 on Build GRCh38
rs1052918301 Chr.2: 27057281-27057281 on Build GRCh38
rs72808950 Chr.2: 27057272-27057272 on Build GRCh38
rs528501838 Chr.2: 27057255-27057255 on Build GRCh38
rs761872429 Chr.2: 27057305-27057305 on Build GRCh38
rs201153256 Chr.2: 27057263-27057263 on Build GRCh38
rs938406761 Chr.2: 27057122-27057122 on Build GRCh38
rs923873303 Chr.2: 27057118-27057118 on Build GRCh38

View Full Product Details