Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53492566-53492839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKTIP
Entrez Gene ID
64400
Gene Name
AKT interacting protein
Gene Aliases
FT1, FTS
UniGene
Hs.380897
Gene Chromosome Location
Chr.16: 53491028-53504411 on Build GRCh38
Transcripts
NM_001012398.2 NM_001308325.1 NM_022476.3 XM_005256095.4 XM_005256096.4 XM_005256097.4 XM_005256098.4 XM_017023564.1 XM_017023565.1 XM_017023566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7207617 c.730T>C p.S244P Chr.16: 53492734-53492734 on Build GRCh38
236973 c.711-3A>G p.? Chr.16: 53492756-53492756 on Build GRCh38
3510250 c.733G>T p.V245L Chr.16: 53492731-53492731 on Build GRCh38
4846515 c.718C>G p.P240A Chr.16: 53492746-53492746 on Build GRCh38
3937092 c.736C>T p.H246Y Chr.16: 53492728-53492728 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540691372 Chr.16: 53492757-53492757 on Build GRCh38
rs16952274 Chr.16: 53492596-53492596 on Build GRCh38
rs372664127 Chr.16: 53492783-53492783 on Build GRCh38
rs764283521 Chr.16: 53492804-53492804 on Build GRCh38
rs796852085 Chr.16: 53492807-53492808 on Build GRCh38
rs756190685 Chr.16: 53492686-53492687 on Build GRCh38
rs138086580 Chr.16: 53492678-53492678 on Build GRCh38
rs780542090 Chr.16: 53492738-53492738 on Build GRCh38
rs752878707 Chr.16: 53492659-53492660 on Build GRCh38
rs57994071 Chr.16: 53492805-53492806 on Build GRCh38
rs201339403 Chr.16: 53492660-53492663 on Build GRCh38
rs751866927 Chr.16: 53492684-53492684 on Build GRCh38
rs750806890 Chr.16: 53492725-53492725 on Build GRCh38
rs749751888 Chr.16: 53492744-53492744 on Build GRCh38
rs762636791 Chr.16: 53492646-53492647 on Build GRCh38
rs549374435 Chr.16: 53492806-53492806 on Build GRCh38
rs200920195 Chr.16: 53492729-53492729 on Build GRCh38
rs745674643 Chr.16: 53492781-53492781 on Build GRCh38
rs374331945 Chr.16: 53492646-53492646 on Build GRCh38
rs765225998 Chr.16: 53492669-53492669 on Build GRCh38
rs767877058 Chr.16: 53492727-53492727 on Build GRCh38
rs781351209 Chr.16: 53492720-53492720 on Build GRCh38
rs11075977 Chr.16: 53492790-53492790 on Build GRCh38
rs757438695 Chr.16: 53492687-53492687 on Build GRCh38
rs367837944 Chr.16: 53492778-53492778 on Build GRCh38
rs777772866 Chr.16: 53492757-53492760 on Build GRCh38
rs370676002 Chr.16: 53492663-53492663 on Build GRCh38
rs753857027 Chr.16: 53492760-53492764 on Build GRCh38
rs971480099 Chr.16: 53492620-53492620 on Build GRCh38
rs374819167 Chr.16: 53492745-53492745 on Build GRCh38
rs9931702 Chr.16: 53492639-53492639 on Build GRCh38
rs770741588 Chr.16: 53492770-53492770 on Build GRCh38
rs762787958 Chr.16: 53492796-53492796 on Build GRCh38

View Full Product Details