Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 100892842-100893115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A3
Entrez Gene ID
220
Gene Name
aldehyde dehydrogenase 1 family member A3
Gene Aliases
ALDH1A6, ALDH6, MCOP8, RALDH3
UniGene
Hs.459538
Gene Chromosome Location
Chr.15: 100879692-100916626 on Build GRCh38
Transcripts
NM_000693.3 NM_001293815.1 NR_135827.1

Gene Symbol
LOC101927751
Entrez Gene ID
101927751
Gene Name
uncharacterized LOC101927751
Gene Aliases
-
UniGene
Hs.659224
Gene Chromosome Location
Chr.15: 100892343-100919283 on Build GRCh38
Transcripts
NM_000693.3 NM_001293815.1 NR_135827.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4136264 c.482A>G p.N161S Chr.15: 100892951-100892951 on Build GRCh38
3815660 c.486C>T p.V162V Chr.15: 100892955-100892955 on Build GRCh38
7191177 c.519C>T p.V173V Chr.15: 100892988-100892988 on Build GRCh38
554357 c.493T>A p.F165I Chr.15: 100892962-100892962 on Build GRCh38
7210470 c.527C>T p.A176V Chr.15: 100892996-100892996 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs961562891 Chr.15: 100892938-100892938 on Build GRCh38
rs761177304 Chr.15: 100892898-100892898 on Build GRCh38
rs753948027 Chr.15: 100892946-100892946 on Build GRCh38
rs577964072 Chr.15: 100893087-100893087 on Build GRCh38
rs767531940 Chr.15: 100893032-100893032 on Build GRCh38
rs199635593 Chr.15: 100893050-100893050 on Build GRCh38
rs567221959 Chr.15: 100892869-100892869 on Build GRCh38
rs748353857 Chr.15: 100892953-100892953 on Build GRCh38
rs761893946 Chr.15: 100893017-100893017 on Build GRCh38
rs370738952 Chr.15: 100893049-100893049 on Build GRCh38
rs776156229 Chr.15: 100892972-100892972 on Build GRCh38
rs778994919 Chr.15: 100893042-100893042 on Build GRCh38
rs755132966 Chr.15: 100893040-100893040 on Build GRCh38
rs754108631 Chr.15: 100892934-100892935 on Build GRCh38
rs763887960 Chr.15: 100892933-100892933 on Build GRCh38
rs759075641 Chr.15: 100892973-100892973 on Build GRCh38
rs779308506 Chr.15: 100892952-100892952 on Build GRCh38
rs764017616 Chr.15: 100892921-100892921 on Build GRCh38
rs763866352 Chr.15: 100892981-100892981 on Build GRCh38
rs764836129 Chr.15: 100892909-100892909 on Build GRCh38
rs573473646 Chr.15: 100893070-100893070 on Build GRCh38
rs538933495 Chr.15: 100892954-100892954 on Build GRCh38
rs954265765 Chr.15: 100892876-100892876 on Build GRCh38
rs562733349 Chr.15: 100893066-100893066 on Build GRCh38
rs1018958822 Chr.15: 100893084-100893084 on Build GRCh38
rs769240976 Chr.15: 100892977-100892977 on Build GRCh38
rs200440536 Chr.15: 100893011-100893011 on Build GRCh38
rs759235011 Chr.15: 100892907-100892907 on Build GRCh38
rs762603642 Chr.15: 100892979-100892979 on Build GRCh38
rs113869203 Chr.15: 100892950-100892950 on Build GRCh38
rs757457119 Chr.15: 100892980-100892981 on Build GRCh38
rs374254045 Chr.15: 100893038-100893038 on Build GRCh38
rs111640914 Chr.15: 100893051-100893051 on Build GRCh38
rs773615296 Chr.15: 100892897-100892897 on Build GRCh38
rs771404117 Chr.15: 100892967-100892967 on Build GRCh38
rs571610912 Chr.15: 100892941-100892941 on Build GRCh38
rs150542846 Chr.15: 100892956-100892956 on Build GRCh38
rs79200242 Chr.15: 100893048-100893048 on Build GRCh38
rs118116058 Chr.15: 100892924-100892924 on Build GRCh38
rs751460261 Chr.15: 100892935-100892935 on Build GRCh38
rs772326662 Chr.15: 100892955-100892955 on Build GRCh38
rs79145913 Chr.15: 100893059-100893059 on Build GRCh38
rs776103489 Chr.15: 100892906-100892906 on Build GRCh38
rs752352897 Chr.15: 100892913-100892913 on Build GRCh38
rs117325892 Chr.15: 100892903-100892903 on Build GRCh38

View Full Product Details