Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24319257-24319486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY4
Entrez Gene ID
196883
Gene Name
adenylate cyclase 4
Gene Aliases
AC4
UniGene
Hs.443428
Gene Chromosome Location
Chr.14: 24318349-24335071 on Build GRCh38
Transcripts
NM_001198568.1 NM_001198592.1 NM_139247.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
955031 c.2782G>A p.G928S Chr.14: 24319388-24319388 on Build GRCh38
4050255 c.2750A>G p.K917R Chr.14: 24319420-24319420 on Build GRCh38
3814631 c.2739C>A p.L913L Chr.14: 24319431-24319431 on Build GRCh38
955032 c.2734-2A>G p.? Chr.14: 24319438-24319438 on Build GRCh38
5743641 c.2747C>T p.P916L Chr.14: 24319423-24319423 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747313455 Chr.14: 24319448-24319448 on Build GRCh38
rs370319489 Chr.14: 24319336-24319336 on Build GRCh38
rs74417323 Chr.14: 24319324-24319324 on Build GRCh38
rs369368195 Chr.14: 24319366-24319366 on Build GRCh38
rs752376679 Chr.14: 24319291-24319291 on Build GRCh38
rs763585938 Chr.14: 24319405-24319405 on Build GRCh38
rs751378155 Chr.14: 24319288-24319288 on Build GRCh38
rs991489441 Chr.14: 24319394-24319394 on Build GRCh38
rs779281281 Chr.14: 24319318-24319318 on Build GRCh38
rs765821696 Chr.14: 24319420-24319420 on Build GRCh38
rs866540700 Chr.14: 24319329-24319329 on Build GRCh38
rs774420099 Chr.14: 24319365-24319365 on Build GRCh38
rs776789122 Chr.14: 24319357-24319357 on Build GRCh38
rs150756728 Chr.14: 24319388-24319388 on Build GRCh38
rs561457351 Chr.14: 24319286-24319286 on Build GRCh38
rs753454736 Chr.14: 24319409-24319409 on Build GRCh38
rs1017948479 Chr.14: 24319306-24319306 on Build GRCh38
rs373827945 Chr.14: 24319359-24319359 on Build GRCh38
rs750896425 Chr.14: 24319422-24319422 on Build GRCh38
rs2295302 Chr.14: 24319325-24319325 on Build GRCh38
rs755725587 Chr.14: 24319295-24319295 on Build GRCh38
rs775385357 Chr.14: 24319389-24319389 on Build GRCh38
rs755235652 Chr.14: 24319451-24319451 on Build GRCh38
rs777375448 Chr.14: 24319298-24319298 on Build GRCh38
rs375846890 Chr.14: 24319350-24319350 on Build GRCh38
rs758860952 Chr.14: 24319434-24319434 on Build GRCh38
rs756706783 Chr.14: 24319410-24319410 on Build GRCh38
rs914579219 Chr.14: 24319443-24319443 on Build GRCh38
rs371596967 Chr.14: 24319445-24319445 on Build GRCh38
rs78220825 Chr.14: 24319395-24319395 on Build GRCh38
rs80151064 Chr.14: 24319317-24319317 on Build GRCh38
rs372403848 Chr.14: 24319363-24319363 on Build GRCh38
rs982000290 Chr.14: 24319345-24319345 on Build GRCh38
rs370012024 Chr.14: 24319313-24319313 on Build GRCh38
rs759403321 Chr.14: 24319290-24319290 on Build GRCh38
rs375047247 Chr.14: 24319453-24319453 on Build GRCh38

View Full Product Details