Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110136585-110136850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP2
Entrez Gene ID
11217
Gene Name
A-kinase anchoring protein 2
Gene Aliases
AKAP-2, AKAPKL, MISP2, PRKA2
UniGene
Hs.591908
Gene Chromosome Location
Chr.9: 110048598-110172512 on Build GRCh38
Transcripts
NM_001004065.4 NM_001136562.2 NM_001198656.1 NM_007203.4 NM_147150.2

Gene Symbol
PALM2-AKAP2
Entrez Gene ID
445815
Gene Name
PALM2-AKAP2 readthrough
Gene Aliases
AKAP2
UniGene
Hs.591908
Gene Chromosome Location
Chr.9: 109780297-110172512 on Build GRCh38
Transcripts
NM_001004065.4 NM_001136562.2 NM_001198656.1 NM_007203.4 NM_147150.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1103791 c.665G>A p.G222D Chr.9: 110136635-110136635 on Build GRCh38
5410118 c.853G>A p.D285N Chr.9: 110136823-110136823 on Build GRCh38
3847307 c.1231G>C p.E411Q Chr.9: 110136775-110136775 on Build GRCh38
6259251 c.1297G>T p.E433* Chr.9: 110136841-110136841 on Build GRCh38
6737113 c.692C>T p.A231V Chr.9: 110136662-110136662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766414145 Chr.9: 110136663-110136663 on Build GRCh38
rs768510991 Chr.9: 110136750-110136750 on Build GRCh38
rs748808900 Chr.9: 110136618-110136618 on Build GRCh38
rs146218630 Chr.9: 110136715-110136715 on Build GRCh38
rs576854132 Chr.9: 110136694-110136694 on Build GRCh38
rs146014470 Chr.9: 110136763-110136763 on Build GRCh38
rs148195165 Chr.9: 110136673-110136673 on Build GRCh38
rs760511732 Chr.9: 110136644-110136644 on Build GRCh38
rs561250097 Chr.9: 110136762-110136762 on Build GRCh38
rs753458424 Chr.9: 110136784-110136784 on Build GRCh38
rs141137319 Chr.9: 110136702-110136702 on Build GRCh38
rs746855386 Chr.9: 110136748-110136748 on Build GRCh38
rs912577574 Chr.9: 110136713-110136713 on Build GRCh38
rs767981419 Chr.9: 110136689-110136689 on Build GRCh38
rs778395434 Chr.9: 110136787-110136787 on Build GRCh38
rs774529525 Chr.9: 110136767-110136767 on Build GRCh38
rs756478584 Chr.9: 110136782-110136782 on Build GRCh38
rs750076648 Chr.9: 110136742-110136742 on Build GRCh38
rs147833425 Chr.9: 110136716-110136716 on Build GRCh38
rs768071507 Chr.9: 110136773-110136773 on Build GRCh38
rs755464316 Chr.9: 110136682-110136682 on Build GRCh38
rs753146690 Chr.9: 110136781-110136781 on Build GRCh38
rs777418174 Chr.9: 110136703-110136703 on Build GRCh38
rs771789071 Chr.9: 110136638-110136638 on Build GRCh38
rs751035084 Chr.9: 110136714-110136714 on Build GRCh38
rs115538409 Chr.9: 110136754-110136754 on Build GRCh38
rs764600135 Chr.9: 110136655-110136655 on Build GRCh38
rs918021457 Chr.9: 110136686-110136686 on Build GRCh38
rs565115211 Chr.9: 110136625-110136625 on Build GRCh38
rs759481374 Chr.9: 110136634-110136634 on Build GRCh38
rs866697999 Chr.9: 110136790-110136790 on Build GRCh38
rs376721653 Chr.9: 110136728-110136728 on Build GRCh38
rs770419829 Chr.9: 110136619-110136619 on Build GRCh38
rs756987750 Chr.9: 110136785-110136785 on Build GRCh38
rs765619092 Chr.9: 110136658-110136658 on Build GRCh38
rs775470940 Chr.9: 110136643-110136643 on Build GRCh38
rs1045395665 Chr.9: 110136777-110136777 on Build GRCh38
rs772172252 Chr.9: 110136741-110136741 on Build GRCh38
rs778441291 Chr.9: 110136726-110136726 on Build GRCh38
rs745447689 Chr.9: 110136824-110136824 on Build GRCh38
rs759531024 Chr.9: 110136770-110136770 on Build GRCh38
rs143371794 Chr.9: 110136662-110136662 on Build GRCh38
rs945679709 Chr.9: 110136755-110136755 on Build GRCh38

View Full Product Details