Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 22978261-22978528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AJUBA
Entrez Gene ID
84962
Gene Name
ajuba LIM protein
Gene Aliases
JUB
UniGene
Hs.655832
Gene Chromosome Location
Chr.14: 22971174-22982642 on Build GRCh38
Transcripts
NM_032876.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1285636 c.1058A>C p.Q353P Chr.14: 22978394-22978394 on Build GRCh38
5436166 c.1013G>A p.C338Y Chr.14: 22978439-22978439 on Build GRCh38
5688437 c.1108+1_1108+3delGTG p.? Chr.14: 22978341-22978343 on Build GRCh38
4432042 c.1007-7_1007-3delTTCTC p.? Chr.14: 22978448-22978452 on Build GRCh38
4977178 c.1057C>T p.Q353* Chr.14: 22978395-22978395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76959995 Chr.14: 22978385-22978385 on Build GRCh38
rs762549983 Chr.14: 22978444-22978444 on Build GRCh38
rs781025877 Chr.14: 22978389-22978389 on Build GRCh38
rs773985579 Chr.14: 22978470-22978470 on Build GRCh38
rs748054763 Chr.14: 22978399-22978399 on Build GRCh38
rs753038177 Chr.14: 22978487-22978487 on Build GRCh38
rs772890059 Chr.14: 22978437-22978437 on Build GRCh38
rs755876832 Chr.14: 22978448-22978452 on Build GRCh38
rs770550047 Chr.14: 22978462-22978462 on Build GRCh38
rs757539118 Chr.14: 22978359-22978359 on Build GRCh38
rs753564914 Chr.14: 22978448-22978448 on Build GRCh38
rs938775828 Chr.14: 22978480-22978480 on Build GRCh38
rs372799493 Chr.14: 22978318-22978318 on Build GRCh38
rs768434215 Chr.14: 22978384-22978384 on Build GRCh38
rs113563394 Chr.14: 22978447-22978447 on Build GRCh38
rs769832788 Chr.14: 22978426-22978426 on Build GRCh38
rs4318130 Chr.14: 22978467-22978467 on Build GRCh38
rs181735426 Chr.14: 22978492-22978492 on Build GRCh38
rs759847978 Chr.14: 22978477-22978477 on Build GRCh38
rs186366443 Chr.14: 22978500-22978500 on Build GRCh38
rs951712053 Chr.14: 22978429-22978429 on Build GRCh38
rs746795497 Chr.14: 22978375-22978375 on Build GRCh38
rs576029085 Chr.14: 22978372-22978372 on Build GRCh38
rs920199806 Chr.14: 22978329-22978329 on Build GRCh38
rs888923113 Chr.14: 22978464-22978464 on Build GRCh38
rs373877597 Chr.14: 22978383-22978384 on Build GRCh38
rs371782085 Chr.14: 22978299-22978299 on Build GRCh38
rs376619101 Chr.14: 22978303-22978303 on Build GRCh38
rs577069427 Chr.14: 22978296-22978296 on Build GRCh38
rs76039198 Chr.14: 22978304-22978304 on Build GRCh38
rs138729157 Chr.14: 22978348-22978348 on Build GRCh38
rs777875023 Chr.14: 22978341-22978341 on Build GRCh38
rs767913361 Chr.14: 22978298-22978298 on Build GRCh38

View Full Product Details