Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38709262-38709535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN4
Entrez Gene ID
81
Gene Name
actinin alpha 4
Gene Aliases
ACTININ-4, FSGS, FSGS1
UniGene
Hs.270291
Gene Chromosome Location
Chr.19: 38647616-38730532 on Build GRCh38
Transcripts
NM_001322033.1 NM_004924.5 XM_005259281.4 XM_006723406.2 XM_017027331.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6593061 c.685G>A p.E229K Chr.19: 38709428-38709428 on Build GRCh38
6593053 c.732G>T p.E244D Chr.19: 38709475-38709475 on Build GRCh38
3938091 c.705C>T p.L235L Chr.19: 38709448-38709448 on Build GRCh38
3222869 c.654C>T p.D218D Chr.19: 38709397-38709397 on Build GRCh38
5587118 c.712C>T p.P238S Chr.19: 38709455-38709455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575966776 Chr.19: 38709397-38709397 on Build GRCh38
rs763051766 Chr.19: 38709428-38709428 on Build GRCh38
rs369348159 Chr.19: 38709376-38709376 on Build GRCh38
rs999373899 Chr.19: 38709446-38709446 on Build GRCh38
rs201242046 Chr.19: 38709372-38709372 on Build GRCh38
rs963855862 Chr.19: 38709442-38709442 on Build GRCh38
rs201961987 Chr.19: 38709488-38709488 on Build GRCh38
rs763760070 Chr.19: 38709427-38709427 on Build GRCh38
rs776682368 Chr.19: 38709388-38709388 on Build GRCh38
rs753037562 Chr.19: 38709499-38709499 on Build GRCh38
rs972933505 Chr.19: 38709308-38709308 on Build GRCh38
rs767053146 Chr.19: 38709464-38709464 on Build GRCh38
rs777391225 Chr.19: 38709485-38709489 on Build GRCh38
rs754339114 Chr.19: 38709466-38709466 on Build GRCh38
rs374419676 Chr.19: 38709349-38709349 on Build GRCh38
rs755403506 Chr.19: 38709368-38709370 on Build GRCh38
rs149399105 Chr.19: 38709487-38709487 on Build GRCh38
rs58197765 Chr.19: 38709361-38709361 on Build GRCh38
rs754173500 Chr.19: 38709354-38709354 on Build GRCh38
rs755305386 Chr.19: 38709356-38709356 on Build GRCh38
rs908187005 Chr.19: 38709306-38709306 on Build GRCh38
rs372891206 Chr.19: 38709360-38709360 on Build GRCh38
rs866990918 Chr.19: 38709430-38709430 on Build GRCh38
rs756747606 Chr.19: 38709345-38709345 on Build GRCh38
rs112958318 Chr.19: 38709478-38709478 on Build GRCh38
rs113222869 Chr.19: 38709477-38709477 on Build GRCh38
rs182290573 Chr.19: 38709290-38709290 on Build GRCh38
rs753371558 Chr.19: 38709505-38709505 on Build GRCh38
rs975304766 Chr.19: 38709288-38709288 on Build GRCh38
rs572926291 Chr.19: 38709303-38709303 on Build GRCh38
rs758713589 Chr.19: 38709502-38709502 on Build GRCh38
rs201630706 Chr.19: 38709392-38709392 on Build GRCh38
rs371499695 Chr.19: 38709341-38709341 on Build GRCh38
rs762661647 Chr.19: 38709420-38709420 on Build GRCh38
rs751192403 Chr.19: 38709448-38709448 on Build GRCh38
rs756768920 Chr.19: 38709457-38709457 on Build GRCh38
rs1038119591 Chr.19: 38709337-38709337 on Build GRCh38
rs757590439 Chr.19: 38709301-38709301 on Build GRCh38
rs775368836 Chr.19: 38709415-38709415 on Build GRCh38
rs749842149 Chr.19: 38709354-38709354 on Build GRCh38
rs1033018062 Chr.19: 38709408-38709408 on Build GRCh38
rs1029555945 Chr.19: 38709480-38709480 on Build GRCh38
rs982834321 Chr.19: 38709304-38709304 on Build GRCh38
rs45561939 Chr.19: 38709358-38709358 on Build GRCh38
rs749414417 Chr.19: 38709500-38709500 on Build GRCh38
rs985541890 Chr.19: 38709286-38709286 on Build GRCh38
rs778036163 Chr.19: 38709513-38709513 on Build GRCh38
rs56737532 Chr.19: 38709340-38709340 on Build GRCh38

View Full Product Details