Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 229525875-229526114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB10
Entrez Gene ID
23456
Gene Name
ATP binding cassette subfamily B member 10
Gene Aliases
EST20237, M-ABC2, MTABC2
UniGene
Hs.17614
Gene Chromosome Location
Chr.1: 229516582-229558987 on Build GRCh38
Transcripts
NM_012089.2 XM_011544135.2 XM_011544136.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235715 c.1841C>T p.A614V Chr.1: 229526001-229526001 on Build GRCh38
7122934 c.1756G>C p.E586Q Chr.1: 229526086-229526086 on Build GRCh38
5597976 c.1849C>T p.R617W Chr.1: 229525993-229525993 on Build GRCh38
6235593 c.1845C>G p.F615L Chr.1: 229525997-229525997 on Build GRCh38
6589962 c.1889delA p.K630fs*20 Chr.1: 229525953-229525953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753294026 Chr.1: 229526045-229526045 on Build GRCh38
rs758127052 Chr.1: 229525979-229525979 on Build GRCh38
rs779835513 Chr.1: 229525993-229525993 on Build GRCh38
rs774717532 Chr.1: 229526029-229526029 on Build GRCh38
rs916144143 Chr.1: 229525983-229525983 on Build GRCh38
rs768094428 Chr.1: 229526044-229526044 on Build GRCh38
rs756855560 Chr.1: 229525970-229525970 on Build GRCh38
rs369960601 Chr.1: 229525952-229525952 on Build GRCh38
rs933101692 Chr.1: 229525992-229525992 on Build GRCh38
rs186898100 Chr.1: 229525994-229525994 on Build GRCh38
rs759998738 Chr.1: 229526031-229526031 on Build GRCh38
rs758014791 Chr.1: 229526073-229526073 on Build GRCh38
rs748196983 Chr.1: 229526006-229526006 on Build GRCh38
rs766824942 Chr.1: 229525942-229525942 on Build GRCh38
rs766513739 Chr.1: 229526022-229526022 on Build GRCh38
rs765405410 Chr.1: 229525909-229525909 on Build GRCh38
rs756802143 Chr.1: 229526047-229526047 on Build GRCh38
rs1037150968 Chr.1: 229526001-229526001 on Build GRCh38
rs776583118 Chr.1: 229525999-229525999 on Build GRCh38
rs182613011 Chr.1: 229525957-229525957 on Build GRCh38
rs375446207 Chr.1: 229525921-229525921 on Build GRCh38
rs746768406 Chr.1: 229526084-229526084 on Build GRCh38
rs773523920 Chr.1: 229525914-229525914 on Build GRCh38
rs752004959 Chr.1: 229525948-229525948 on Build GRCh38
rs563275826 Chr.1: 229526010-229526010 on Build GRCh38
rs749910327 Chr.1: 229526071-229526071 on Build GRCh38
rs373067837 Chr.1: 229525977-229525977 on Build GRCh38
rs532293063 Chr.1: 229526014-229526014 on Build GRCh38
rs763301962 Chr.1: 229526016-229526016 on Build GRCh38
rs749794068 Chr.1: 229525976-229525976 on Build GRCh38
rs1004343437 Chr.1: 229525996-229525996 on Build GRCh38
rs898786706 Chr.1: 229526017-229526017 on Build GRCh38
rs1022993774 Chr.1: 229526059-229526059 on Build GRCh38
rs779598408 Chr.1: 229526080-229526080 on Build GRCh38
rs753347027 Chr.1: 229525958-229525958 on Build GRCh38
rs778119051 Chr.1: 229525964-229525965 on Build GRCh38
rs764872659 Chr.1: 229526061-229526061 on Build GRCh38

View Full Product Details