Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122077701-122077962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AASS
Entrez Gene ID
10157
Gene Name
aminoadipate-semialdehyde synthase
Gene Aliases
LKR/SDH, LKRSDH, LORSDH
UniGene
Hs.156738
Gene Chromosome Location
Chr.7: 122073544-122144290 on Build GRCh38
Transcripts
NM_005763.3 XM_011515725.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4153790 c.2620G>A p.V874M Chr.7: 122077880-122077880 on Build GRCh38
598864 c.2565G>A p.T855T Chr.7: 122077935-122077935 on Build GRCh38
4168145 c.2564C>T p.T855M Chr.7: 122077936-122077936 on Build GRCh38
3995323 c.2584G>T p.G862W Chr.7: 122077916-122077916 on Build GRCh38
3995324 c.2583T>C p.Y861Y Chr.7: 122077917-122077917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147476318 Chr.7: 122077865-122077866 on Build GRCh38
rs752702037 Chr.7: 122077904-122077904 on Build GRCh38
rs760663934 Chr.7: 122077808-122077808 on Build GRCh38
rs972977767 Chr.7: 122077828-122077828 on Build GRCh38
rs1003600021 Chr.7: 122077781-122077781 on Build GRCh38
rs943946020 Chr.7: 122077881-122077881 on Build GRCh38
rs150593459 Chr.7: 122077758-122077758 on Build GRCh38
rs768298788 Chr.7: 122077866-122077866 on Build GRCh38
rs961040266 Chr.7: 122077766-122077766 on Build GRCh38
rs779193257 Chr.7: 122077747-122077747 on Build GRCh38
rs531058371 Chr.7: 122077750-122077750 on Build GRCh38
rs769437600 Chr.7: 122077740-122077740 on Build GRCh38
rs993503401 Chr.7: 122077751-122077751 on Build GRCh38
rs116061308 Chr.7: 122077880-122077880 on Build GRCh38
rs905307693 Chr.7: 122077775-122077775 on Build GRCh38
rs777824271 Chr.7: 122077917-122077917 on Build GRCh38
rs891766148 Chr.7: 122077929-122077929 on Build GRCh38
rs747351819 Chr.7: 122077845-122077845 on Build GRCh38
rs746330018 Chr.7: 122077865-122077865 on Build GRCh38
rs587777121 Chr.7: 122077833-122077837 on Build GRCh38
rs766283847 Chr.7: 122077809-122077809 on Build GRCh38
rs760432920 Chr.7: 122077872-122077872 on Build GRCh38
rs750181686 Chr.7: 122077796-122077796 on Build GRCh38
rs1034958206 Chr.7: 122077743-122077743 on Build GRCh38
rs530285419 Chr.7: 122077862-122077862 on Build GRCh38
rs773110350 Chr.7: 122077870-122077870 on Build GRCh38
rs149105850 Chr.7: 122077868-122077869 on Build GRCh38
rs367958255 Chr.7: 122077786-122077786 on Build GRCh38
rs1025596306 Chr.7: 122077833-122077833 on Build GRCh38
rs972426537 Chr.7: 122077778-122077778 on Build GRCh38
rs766111824 Chr.7: 122077875-122077875 on Build GRCh38
rs778255100 Chr.7: 122077841-122077841 on Build GRCh38
rs778798520 Chr.7: 122077818-122077818 on Build GRCh38
rs751788580 Chr.7: 122077928-122077928 on Build GRCh38
rs771603825 Chr.7: 122077850-122077850 on Build GRCh38
rs759400940 Chr.7: 122077891-122077891 on Build GRCh38
rs771610987 Chr.7: 122077868-122077868 on Build GRCh38
rs565209129 Chr.7: 122077727-122077727 on Build GRCh38
rs138726205 Chr.7: 122077906-122077906 on Build GRCh38
rs752968291 Chr.7: 122077821-122077821 on Build GRCh38
rs765424237 Chr.7: 122077895-122077895 on Build GRCh38
rs1001709486 Chr.7: 122077739-122077739 on Build GRCh38
rs373242380 Chr.7: 122077838-122077838 on Build GRCh38
rs761594354 Chr.7: 122077867-122077867 on Build GRCh38
rs371822556 Chr.7: 122077810-122077810 on Build GRCh38

View Full Product Details