Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89133069-89133330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF3
Entrez Gene ID
197322
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
-
UniGene
Hs.461727
Gene Chromosome Location
Chr.16: 89093809-89160556 on Build GRCh38
Transcripts
NM_001127214.3 NM_001243279.2 NM_001284316.1 NM_174917.4 NR_104293.1 XM_005256293.2 XM_011522942.1 XM_011522943.1 XM_017023018.1 XM_017023019.1 XM_017023020.1 XM_017023021.1 XM_017023022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3980245 c.1366+5G>A p.? Chr.16: 89133267-89133267 on Build GRCh38
5479239 c.1300C>T p.R434* Chr.16: 89133196-89133196 on Build GRCh38
4976049 c.1246C>T p.P416S Chr.16: 89133142-89133142 on Build GRCh38
2922775 c.1353C>G p.G451G Chr.16: 89133249-89133249 on Build GRCh38
4141953 c.1283G>T p.R428M Chr.16: 89133179-89133179 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137995833 Chr.16: 89133197-89133197 on Build GRCh38
rs745439171 Chr.16: 89133233-89133233 on Build GRCh38
rs3743984 Chr.16: 89133147-89133147 on Build GRCh38
rs759209996 Chr.16: 89133093-89133093 on Build GRCh38
rs768562298 Chr.16: 89133159-89133159 on Build GRCh38
rs1057243097 Chr.16: 89133171-89133171 on Build GRCh38
rs764608253 Chr.16: 89133192-89133192 on Build GRCh38
rs766713747 Chr.16: 89133138-89133138 on Build GRCh38
rs374578722 Chr.16: 89133141-89133141 on Build GRCh38
rs761054207 Chr.16: 89133188-89133188 on Build GRCh38
rs766764090 Chr.16: 89133196-89133196 on Build GRCh38
rs776385863 Chr.16: 89133245-89133245 on Build GRCh38
rs758641886 Chr.16: 89133117-89133117 on Build GRCh38
rs756661716 Chr.16: 89133224-89133224 on Build GRCh38
rs1050796580 Chr.16: 89133218-89133218 on Build GRCh38
rs750555101 Chr.16: 89133174-89133174 on Build GRCh38
rs771701041 Chr.16: 89133146-89133146 on Build GRCh38
rs1022155115 Chr.16: 89133097-89133097 on Build GRCh38
rs751859162 Chr.16: 89133283-89133283 on Build GRCh38
rs766676473 Chr.16: 89133279-89133279 on Build GRCh38
rs142092069 Chr.16: 89133230-89133230 on Build GRCh38
rs554194567 Chr.16: 89133239-89133239 on Build GRCh38
rs756643778 Chr.16: 89133303-89133303 on Build GRCh38
rs372495378 Chr.16: 89133265-89133265 on Build GRCh38
rs763231512 Chr.16: 89133190-89133190 on Build GRCh38
rs745543610 Chr.16: 89133142-89133142 on Build GRCh38
rs538654192 Chr.16: 89133119-89133119 on Build GRCh38
rs775794698 Chr.16: 89133162-89133162 on Build GRCh38
rs768004632 Chr.16: 89133298-89133298 on Build GRCh38
rs1049672981 Chr.16: 89133182-89133182 on Build GRCh38
rs749930101 Chr.16: 89133309-89133309 on Build GRCh38
rs12447947 Chr.16: 89133243-89133243 on Build GRCh38
rs370783227 Chr.16: 89133164-89133164 on Build GRCh38
rs770503655 Chr.16: 89133136-89133136 on Build GRCh38
rs150322170 Chr.16: 89133162-89133162 on Build GRCh38
rs776995095 Chr.16: 89133224-89133225 on Build GRCh38
rs761825392 Chr.16: 89133251-89133251 on Build GRCh38
rs760524726 Chr.16: 89133153-89133153 on Build GRCh38
rs781776532 Chr.16: 89133131-89133131 on Build GRCh38
rs764087633 Chr.16: 89133109-89133109 on Build GRCh38
rs773327286 Chr.16: 89133270-89133270 on Build GRCh38
rs199736898 Chr.16: 89133103-89133103 on Build GRCh38
rs752037142 Chr.16: 89133118-89133118 on Build GRCh38
rs375414491 Chr.16: 89133201-89133201 on Build GRCh38
rs753076046 Chr.16: 89133302-89133302 on Build GRCh38
rs771104684 Chr.16: 89133294-89133294 on Build GRCh38
rs759927049 Chr.16: 89133285-89133285 on Build GRCh38
rs753904828 Chr.16: 89133113-89133113 on Build GRCh38
rs765382177 Chr.16: 89133166-89133166 on Build GRCh38
rs771704373 Chr.16: 89133234-89133234 on Build GRCh38
rs775351320 Chr.16: 89133105-89133105 on Build GRCh38
rs375591399 Chr.16: 89133308-89133308 on Build GRCh38
rs748712754 Chr.16: 89133133-89133133 on Build GRCh38
rs998246075 Chr.16: 89133216-89133216 on Build GRCh38
rs750688940 Chr.16: 89133116-89133116 on Build GRCh38
rs118065221 Chr.16: 89133114-89133114 on Build GRCh38
rs762991611 Chr.16: 89133273-89133273 on Build GRCh38

View Full Product Details