Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244415880-244416153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSS
Entrez Gene ID
159
Gene Name
adenylosuccinate synthase
Gene Aliases
ADEH, ADSS 2
UniGene
Hs.498313
Gene Chromosome Location
Chr.1: 244408492-244452134 on Build GRCh38
Transcripts
NM_001126.3 XM_011544108.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6409472 c.1145G>T p.G382V Chr.1: 244416004-244416004 on Build GRCh38
679594 c.1118A>T p.K373I Chr.1: 244416031-244416031 on Build GRCh38
345020 c.1110G>T p.T370T Chr.1: 244416039-244416039 on Build GRCh38
6260624 c.1166C>A p.P389Q Chr.1: 244415983-244415983 on Build GRCh38
80834 c.1083C>G p.T361T Chr.1: 244416066-244416066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778770860 Chr.1: 244415967-244415967 on Build GRCh38
rs747445427 Chr.1: 244416009-244416009 on Build GRCh38
rs759326155 Chr.1: 244416066-244416066 on Build GRCh38
rs530183884 Chr.1: 244415947-244415947 on Build GRCh38
rs756953317 Chr.1: 244415958-244415958 on Build GRCh38
rs750079350 Chr.1: 244416121-244416121 on Build GRCh38
rs1056400137 Chr.1: 244416106-244416106 on Build GRCh38
rs760664748 Chr.1: 244416083-244416083 on Build GRCh38
rs753445114 Chr.1: 244415942-244415942 on Build GRCh38
rs775394358 Chr.1: 244416078-244416078 on Build GRCh38
rs1036798653 Chr.1: 244416091-244416091 on Build GRCh38
rs761409831 Chr.1: 244416101-244416101 on Build GRCh38
rs764938743 Chr.1: 244416110-244416110 on Build GRCh38
rs764182709 Chr.1: 244416088-244416088 on Build GRCh38
rs773318896 Chr.1: 244416063-244416063 on Build GRCh38
rs377279282 Chr.1: 244416076-244416076 on Build GRCh38
rs750745797 Chr.1: 244415992-244415992 on Build GRCh38
rs1036128904 Chr.1: 244416105-244416105 on Build GRCh38
rs374969795 Chr.1: 244416098-244416098 on Build GRCh38
rs909316747 Chr.1: 244416047-244416047 on Build GRCh38
rs139329929 Chr.1: 244416039-244416039 on Build GRCh38
rs781237785 Chr.1: 244416032-244416032 on Build GRCh38
rs769202132 Chr.1: 244416014-244416014 on Build GRCh38
rs777948763 Chr.1: 244416095-244416095 on Build GRCh38
rs754413969 Chr.1: 244416103-244416104 on Build GRCh38
rs751547117 Chr.1: 244415946-244415948 on Build GRCh38
rs550513045 Chr.1: 244415996-244415996 on Build GRCh38
rs771436487 Chr.1: 244416064-244416064 on Build GRCh38
rs922148883 Chr.1: 244415946-244415946 on Build GRCh38
rs147946123 Chr.1: 244416040-244416040 on Build GRCh38

View Full Product Details