Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 47077330-47077603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMIGO2
Entrez Gene ID
347902
Gene Name
adhesion molecule with Ig like domain 2
Gene Aliases
ALI1, AMIGO-2, DEGA
UniGene
Hs.121520
Gene Chromosome Location
Chr.12: 47075707-47080826 on Build GRCh38
Transcripts
NM_001143668.1 NM_181847.4 XM_005268837.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
939644 c.1433G>A p.G478E Chr.12: 47077570-47077570 on Build GRCh38
939643 c.1476A>C p.I492I Chr.12: 47077527-47077527 on Build GRCh38
3671077 c.1504G>T p.G502W Chr.12: 47077499-47077499 on Build GRCh38
3739677 c.1486A>G p.I496V Chr.12: 47077517-47077517 on Build GRCh38
1746990 c.1477G>C p.A493P Chr.12: 47077526-47077526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774751022 Chr.12: 47077399-47077403 on Build GRCh38
rs751474565 Chr.12: 47077454-47077454 on Build GRCh38
rs754703315 Chr.12: 47077563-47077563 on Build GRCh38
rs773867210 Chr.12: 47077490-47077490 on Build GRCh38
rs551312914 Chr.12: 47077568-47077568 on Build GRCh38
rs986538871 Chr.12: 47077374-47077374 on Build GRCh38
rs757258232 Chr.12: 47077473-47077473 on Build GRCh38
rs768085593 Chr.12: 47077408-47077408 on Build GRCh38
rs754109972 Chr.12: 47077443-47077443 on Build GRCh38
rs761101075 Chr.12: 47077424-47077424 on Build GRCh38
rs368276011 Chr.12: 47077551-47077551 on Build GRCh38
rs755896518 Chr.12: 47077479-47077479 on Build GRCh38
rs772688049 Chr.12: 47077493-47077493 on Build GRCh38
rs368639800 Chr.12: 47077529-47077529 on Build GRCh38
rs944828768 Chr.12: 47077530-47077530 on Build GRCh38
rs755912880 Chr.12: 47077545-47077545 on Build GRCh38
rs780968556 Chr.12: 47077474-47077474 on Build GRCh38
rs201591333 Chr.12: 47077523-47077523 on Build GRCh38
rs773784770 Chr.12: 47077400-47077400 on Build GRCh38
rs912352851 Chr.12: 47077376-47077376 on Build GRCh38
rs370161697 Chr.12: 47077494-47077494 on Build GRCh38
rs11836128 Chr.12: 47077521-47077521 on Build GRCh38
rs779771382 Chr.12: 47077548-47077548 on Build GRCh38
rs766768240 Chr.12: 47077433-47077433 on Build GRCh38
rs139663398 Chr.12: 47077444-47077444 on Build GRCh38
rs765346570 Chr.12: 47077519-47077519 on Build GRCh38
rs761190701 Chr.12: 47077498-47077498 on Build GRCh38
rs745733264 Chr.12: 47077475-47077475 on Build GRCh38
rs145162431 Chr.12: 47077487-47077487 on Build GRCh38
rs745618518 Chr.12: 47077393-47077393 on Build GRCh38
rs374019268 Chr.12: 47077392-47077392 on Build GRCh38
rs547623888 Chr.12: 47077500-47077500 on Build GRCh38
rs140364757 Chr.12: 47077574-47077574 on Build GRCh38
rs748879545 Chr.12: 47077486-47077486 on Build GRCh38
rs1052972535 Chr.12: 47077543-47077543 on Build GRCh38
rs759651263 Chr.12: 47077562-47077564 on Build GRCh38
rs750290860 Chr.12: 47077542-47077542 on Build GRCh38
rs774730267 Chr.12: 47077409-47077409 on Build GRCh38
rs377721693 Chr.12: 47077407-47077407 on Build GRCh38
rs779683406 Chr.12: 47077484-47077484 on Build GRCh38
rs372187703 Chr.12: 47077579-47077579 on Build GRCh38
rs374631836 Chr.12: 47077515-47077515 on Build GRCh38
rs374161995 Chr.12: 47077418-47077418 on Build GRCh38
rs367792195 Chr.12: 47077422-47077422 on Build GRCh38
rs765304150 Chr.12: 47077448-47077448 on Build GRCh38
rs187435585 Chr.12: 47077417-47077417 on Build GRCh38
rs761794586 Chr.12: 47077359-47077359 on Build GRCh38
rs775109262 Chr.12: 47077398-47077398 on Build GRCh38
rs747584427 Chr.12: 47077492-47077492 on Build GRCh38
rs1002414024 Chr.12: 47077368-47077368 on Build GRCh38
rs778411828 Chr.12: 47077564-47077564 on Build GRCh38

View Full Product Details