Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236686667-236686908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN2
Entrez Gene ID
88
Gene Name
actinin alpha 2
Gene Aliases
CMD1AA, CMH23
UniGene
Hs.498178
Gene Chromosome Location
Chr.1: 236686454-236764631 on Build GRCh38
Transcripts
NM_001103.3 NM_001278343.1 NM_001278344.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7110531 c.63C>G p.I21M Chr.1: 236686736-236686736 on Build GRCh38
6429997 c.26A>G p.Q9R Chr.1: 236686699-236686699 on Build GRCh38
4926324 c.85G>A p.D29N Chr.1: 236686758-236686758 on Build GRCh38
4926323 c.85G>A p.D29N Chr.1: 236686758-236686758 on Build GRCh38
6456913 c.61A>C p.I21L Chr.1: 236686734-236686734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006877564 Chr.1: 236686873-236686873 on Build GRCh38
rs397516586 Chr.1: 236686766-236686766 on Build GRCh38
rs886046205 Chr.1: 236686704-236686704 on Build GRCh38
rs1057518609 Chr.1: 236686738-236686738 on Build GRCh38
rs775052416 Chr.1: 236686719-236686719 on Build GRCh38
rs551141480 Chr.1: 236686695-236686695 on Build GRCh38
rs376510300 Chr.1: 236686786-236686786 on Build GRCh38
rs771424965 Chr.1: 236686718-236686718 on Build GRCh38
rs530707458 Chr.1: 236686790-236686790 on Build GRCh38
rs763227511 Chr.1: 236686760-236686760 on Build GRCh38
rs746504347 Chr.1: 236686832-236686832 on Build GRCh38
rs1048525866 Chr.1: 236686853-236686853 on Build GRCh38
rs1013608004 Chr.1: 236686863-236686870 on Build GRCh38
rs750574123 Chr.1: 236686758-236686758 on Build GRCh38
rs1047065829 Chr.1: 236686867-236686867 on Build GRCh38
rs567105469 Chr.1: 236686871-236686871 on Build GRCh38
rs768605809 Chr.1: 236686727-236686727 on Build GRCh38
rs768599300 Chr.1: 236686839-236686839 on Build GRCh38
rs781746567 Chr.1: 236686795-236686795 on Build GRCh38
rs753143220 Chr.1: 236686806-236686806 on Build GRCh38
rs761544833 Chr.1: 236686739-236686739 on Build GRCh38
rs745563404 Chr.1: 236686814-236686814 on Build GRCh38
rs368367224 Chr.1: 236686691-236686691 on Build GRCh38
rs771008376 Chr.1: 236686714-236686716 on Build GRCh38
rs121434525 Chr.1: 236686699-236686699 on Build GRCh38
rs779469857 Chr.1: 236686831-236686831 on Build GRCh38
rs397516580 Chr.1: 236686721-236686721 on Build GRCh38
rs756481602 Chr.1: 236686811-236686811 on Build GRCh38
rs776456711 Chr.1: 236686732-236686732 on Build GRCh38
rs1047184412 Chr.1: 236686828-236686828 on Build GRCh38
rs771748768 Chr.1: 236686825-236686825 on Build GRCh38
rs730880040 Chr.1: 236686755-236686755 on Build GRCh38
rs373269478 Chr.1: 236686813-236686813 on Build GRCh38
rs765471221 Chr.1: 236686752-236686752 on Build GRCh38
rs776507046 Chr.1: 236686845-236686845 on Build GRCh38

View Full Product Details