Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5880237-5880508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD16
Entrez Gene ID
54522
Gene Name
ankyrin repeat domain 16
Gene Aliases
-
UniGene
Hs.289828
Gene Chromosome Location
Chr.10: 5861726-5889897 on Build GRCh38
Transcripts
NM_001009941.2 NM_001009943.2 NM_019046.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1245014 c.923G>A p.R308Q Chr.10: 5880303-5880303 on Build GRCh38
6581685 c.869T>A p.I290N Chr.10: 5880357-5880357 on Build GRCh38
539204 c.926C>G p.S309* Chr.10: 5880300-5880300 on Build GRCh38
4014828 c.877C>A p.L293I Chr.10: 5880349-5880349 on Build GRCh38
174347 c.922C>T p.R308* Chr.10: 5880304-5880304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774806484 Chr.10: 5880304-5880304 on Build GRCh38
rs769429813 Chr.10: 5880369-5880369 on Build GRCh38
rs1002255675 Chr.10: 5880399-5880399 on Build GRCh38
rs569238552 Chr.10: 5880435-5880435 on Build GRCh38
rs373493365 Chr.10: 5880407-5880407 on Build GRCh38
rs777092982 Chr.10: 5880368-5880369 on Build GRCh38
rs896760401 Chr.10: 5880301-5880301 on Build GRCh38
rs942443378 Chr.10: 5880303-5880303 on Build GRCh38
rs141938525 Chr.10: 5880428-5880428 on Build GRCh38
rs777688711 Chr.10: 5880275-5880275 on Build GRCh38
rs555692126 Chr.10: 5880473-5880473 on Build GRCh38
rs1017719402 Chr.10: 5880400-5880400 on Build GRCh38
rs192228502 Chr.10: 5880452-5880452 on Build GRCh38
rs148178134 Chr.10: 5880292-5880292 on Build GRCh38
rs779857733 Chr.10: 5880372-5880372 on Build GRCh38
rs368951227 Chr.10: 5880307-5880307 on Build GRCh38
rs139786252 Chr.10: 5880343-5880343 on Build GRCh38
rs138975719 Chr.10: 5880468-5880468 on Build GRCh38
rs959871454 Chr.10: 5880424-5880424 on Build GRCh38
rs765437974 Chr.10: 5880414-5880414 on Build GRCh38
rs539631362 Chr.10: 5880436-5880436 on Build GRCh38
rs757972825 Chr.10: 5880305-5880306 on Build GRCh38
rs762282461 Chr.10: 5880306-5880306 on Build GRCh38
rs747731553 Chr.10: 5880277-5880278 on Build GRCh38
rs901309070 Chr.10: 5880326-5880326 on Build GRCh38
rs766532561 Chr.10: 5880324-5880324 on Build GRCh38
rs780713389 Chr.10: 5880366-5880366 on Build GRCh38
rs773835539 Chr.10: 5880389-5880389 on Build GRCh38
rs375360123 Chr.10: 5880386-5880386 on Build GRCh38
rs759535065 Chr.10: 5880409-5880409 on Build GRCh38
rs539021095 Chr.10: 5880420-5880420 on Build GRCh38
rs760661235 Chr.10: 5880310-5880310 on Build GRCh38
rs748520828 Chr.10: 5880375-5880375 on Build GRCh38
rs777670684 Chr.10: 5880354-5880354 on Build GRCh38
rs779828349 Chr.10: 5880293-5880293 on Build GRCh38
rs773925326 Chr.10: 5880308-5880308 on Build GRCh38
rs758254300 Chr.10: 5880349-5880349 on Build GRCh38
rs759767191 Chr.10: 5880331-5880331 on Build GRCh38
rs747595388 Chr.10: 5880394-5880394 on Build GRCh38
rs757648721 Chr.10: 5880359-5880359 on Build GRCh38
rs745392150 Chr.10: 5880367-5880367 on Build GRCh38
rs567975932 Chr.10: 5880280-5880280 on Build GRCh38
rs369759752 Chr.10: 5880397-5880397 on Build GRCh38
rs950698690 Chr.10: 5880447-5880447 on Build GRCh38
rs149338217 Chr.10: 5880347-5880347 on Build GRCh38
rs769362719 Chr.10: 5880298-5880298 on Build GRCh38
rs146132104 Chr.10: 5880461-5880461 on Build GRCh38
rs372629789 Chr.10: 5880358-5880358 on Build GRCh38

View Full Product Details