Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36874494-36874767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf89
Entrez Gene ID
221477
Gene Name
chromosome 6 open reading frame 89
Gene Aliases
BRAP
UniGene
Hs.433381
Gene Chromosome Location
Chr.6: 36834886-36928964 on Build GRCh38
Transcripts
NM_001286636.1 XM_017010434.1 NM_016059.4

Gene Symbol
PPIL1
Entrez Gene ID
51645
Gene Name
peptidylprolyl isomerase like 1
Gene Aliases
CGI-124, CYPL1, PPIase, hCyPX
UniGene
Hs.27693
Gene Chromosome Location
Chr.6: 36854829-36875024 on Build GRCh38
Transcripts
NM_001286636.1 XM_017010434.1 NM_016059.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
76029 c.8C>T p.A3V Chr.6: 36874765-36874765 on Build GRCh38
6327175 c.27G>T p.W9C Chr.6: 36874746-36874746 on Build GRCh38
3697413 c.7G>A p.A3T Chr.6: 36874766-36874766 on Build GRCh38
5588712 c.15C>T p.P5P Chr.6: 36874758-36874758 on Build GRCh38
6572023 c.17C>T p.P6L Chr.6: 36874756-36874756 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs532362718 Chr.6: 36874558-36874558 on Build GRCh38
rs986806455 Chr.6: 36874736-36874736 on Build GRCh38
rs192758601 Chr.6: 36874559-36874559 on Build GRCh38
rs889059138 Chr.6: 36874705-36874705 on Build GRCh38
rs202058478 Chr.6: 36874702-36874702 on Build GRCh38
rs72846078 Chr.6: 36874544-36874544 on Build GRCh38
rs1021452162 Chr.6: 36874550-36874550 on Build GRCh38
rs374199616 Chr.6: 36874735-36874735 on Build GRCh38
rs957667688 Chr.6: 36874623-36874623 on Build GRCh38
rs754069371 Chr.6: 36874693-36874693 on Build GRCh38
rs200226282 Chr.6: 36874696-36874696 on Build GRCh38
rs940828503 Chr.6: 36874639-36874639 on Build GRCh38
rs751349934 Chr.6: 36874677-36874677 on Build GRCh38
rs960740108 Chr.6: 36874580-36874580 on Build GRCh38
rs764069487 Chr.6: 36874673-36874673 on Build GRCh38
rs761591768 Chr.6: 36874678-36874678 on Build GRCh38
rs909383547 Chr.6: 36874626-36874626 on Build GRCh38
rs543327606 Chr.6: 36874542-36874542 on Build GRCh38
rs757713581 Chr.6: 36874741-36874741 on Build GRCh38
rs372188657 Chr.6: 36874573-36874573 on Build GRCh38
rs565804888 Chr.6: 36874562-36874562 on Build GRCh38
rs376803178 Chr.6: 36874576-36874576 on Build GRCh38
rs916850132 Chr.6: 36874597-36874597 on Build GRCh38
rs2303993 Chr.6: 36874651-36874651 on Build GRCh38
rs531569699 Chr.6: 36874541-36874541 on Build GRCh38
rs372066813 Chr.6: 36874698-36874698 on Build GRCh38
rs371206519 Chr.6: 36874668-36874668 on Build GRCh38
rs374192648 Chr.6: 36874694-36874694 on Build GRCh38
rs145636632 Chr.6: 36874653-36874653 on Build GRCh38
rs758871036 Chr.6: 36874700-36874700 on Build GRCh38
rs990280495 Chr.6: 36874637-36874637 on Build GRCh38
rs992543340 Chr.6: 36874590-36874590 on Build GRCh38
rs1031506611 Chr.6: 36874582-36874582 on Build GRCh38
rs548262317 Chr.6: 36874609-36874609 on Build GRCh38
rs781636154 Chr.6: 36874747-36874747 on Build GRCh38
rs371124969 Chr.6: 36874587-36874587 on Build GRCh38

View Full Product Details