Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67438755-67438984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0D1
Entrez Gene ID
9114
Gene Name
ATPase H+ transporting V0 subunit d1
Gene Aliases
ATP6D, ATP6DV, P39, VATX, VMA6, VPATPD
UniGene
Hs.106876
Gene Chromosome Location
Chr.16: 67438014-67481186 on Build GRCh38
Transcripts
NM_004691.4 XM_011523444.1 XM_011523445.1 XM_011523446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1184117 c.882C>A p.F294L Chr.16: 67438805-67438805 on Build GRCh38
6608068 c.817-8delC p.? Chr.16: 67438878-67438878 on Build GRCh38
6608076 c.816G>A p.P272P Chr.16: 67438971-67438971 on Build GRCh38
5054994 c.872A>C p.E291A Chr.16: 67438815-67438815 on Build GRCh38
7212104 c.835G>A p.E279K Chr.16: 67438852-67438852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72650138 Chr.16: 67438919-67438919 on Build GRCh38
rs112420676 Chr.16: 67438790-67438790 on Build GRCh38
rs765114517 Chr.16: 67438805-67438805 on Build GRCh38
rs759472919 Chr.16: 67438789-67438789 on Build GRCh38
rs964215939 Chr.16: 67438837-67438837 on Build GRCh38
rs749567809 Chr.16: 67438879-67438879 on Build GRCh38
rs72650136 Chr.16: 67438917-67438917 on Build GRCh38
rs767020393 Chr.16: 67438935-67438935 on Build GRCh38
rs772264038 Chr.16: 67438887-67438887 on Build GRCh38
rs758364505 Chr.16: 67438822-67438822 on Build GRCh38
rs779642042 Chr.16: 67438949-67438949 on Build GRCh38
rs111951991 Chr.16: 67438872-67438872 on Build GRCh38
rs549838985 Chr.16: 67438777-67438777 on Build GRCh38
rs754268417 Chr.16: 67438950-67438950 on Build GRCh38
rs1024330466 Chr.16: 67438889-67438889 on Build GRCh38
rs780401144 Chr.16: 67438862-67438862 on Build GRCh38
rs72650135 Chr.16: 67438881-67438881 on Build GRCh38
rs768234082 Chr.16: 67438953-67438953 on Build GRCh38
rs72650139 Chr.16: 67438959-67438959 on Build GRCh38
rs201131049 Chr.16: 67438852-67438852 on Build GRCh38
rs201883077 Chr.16: 67438775-67438775 on Build GRCh38
rs773633139 Chr.16: 67438890-67438890 on Build GRCh38
rs751126496 Chr.16: 67438838-67438838 on Build GRCh38
rs376498424 Chr.16: 67438944-67438944 on Build GRCh38
rs777899095 Chr.16: 67438884-67438885 on Build GRCh38
rs748596250 Chr.16: 67438961-67438961 on Build GRCh38
rs752206845 Chr.16: 67438877-67438877 on Build GRCh38
rs66719440 Chr.16: 67438885-67438885 on Build GRCh38
rs764144590 Chr.16: 67438926-67438926 on Build GRCh38
rs756152539 Chr.16: 67438877-67438878 on Build GRCh38
rs761381556 Chr.16: 67438934-67438934 on Build GRCh38
rs148076758 Chr.16: 67438853-67438853 on Build GRCh38
rs72650140 Chr.16: 67438960-67438960 on Build GRCh38
rs755703812 Chr.16: 67438858-67438858 on Build GRCh38
rs760980028 Chr.16: 67438893-67438893 on Build GRCh38
rs72650137 Chr.16: 67438918-67438918 on Build GRCh38
rs763884699 Chr.16: 67438836-67438836 on Build GRCh38
rs769237299 Chr.16: 67438884-67438884 on Build GRCh38
rs771215798 Chr.16: 67438915-67438915 on Build GRCh38
rs368770804 Chr.16: 67438945-67438945 on Build GRCh38
rs891886446 Chr.16: 67438868-67438868 on Build GRCh38
rs752486715 Chr.16: 67438814-67438814 on Build GRCh38
rs772638504 Chr.16: 67438962-67438962 on Build GRCh38
rs751441842 Chr.16: 67438931-67438931 on Build GRCh38

View Full Product Details